Skip to Content
Merck
All Photos(1)

Key Documents

E1895

Sigma-Aldrich

Erlose

≥94% (HPLC)

Synonym(s):

α-D-Glc-(1→4)-α-D-Glc-(1→2)-β-D-Fru, α-Maltosyl β-fructofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16
CAS Number:
Molecular Weight:
504.44
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant

Assay

≥94% (HPLC)

form

powder

optical activity

[α]20/D 108.5 to 112.0 °, c = 0.84% (w/v) in water

storage condition

desiccated

impurities

≤12.5% water (Karl Fischer)

color

white to off-white

solubility

H2O: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H32O16/c19-1-5-8(23)10(25)12(27)16(30-5)32-14-7(3-21)31-17(13(28)11(14)26)34-18(4-22)15(29)9(24)6(2-20)33-18/h5-17,19-29H,1-4H2/t5-,6-,7-,8-,9-,10+,11-,12-,13-,14-,15+,16-,17-,18+/m1/s1

InChI key

FVVCFHXLWDDRHG-KKNDGLDKSA-N

Application

Erlose, a triose sugar (trisaccharide), is used in studies on dietary preference and utilization of triose sugars from aphid honeydew by various insects, such as honey bees and ants. Erlose may be used as a reference compound in assays that analyze the sugars of foods such as royal jelly and honey.

Caution

moisture sensitive

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Julian Görl et al.
Chembiochem : a European journal of chemical biology, 13(1), 149-156 (2011-11-30)
An isomelezitose synthase was redesigned out of the sucrose isomerase from Protaminobacter rubrum for the synthesis of isomelezitose (6-O(F)-glucosylsucrose), a potential nutraceutical. The variants F297A, F297P, R333K, F321A_F319A and E428D catalyze the formation of isomelezitose in up to 70 %
Kris A G Wyckhuys et al.
Journal of insect physiology, 54(2), 481-491 (2008-01-15)
Parasitoids commonly forage in agricultural settings where the predominant sugar source is homopteran honeydew. The aphidiine braconid, Binodoxys communis, is an Asian parasitoid currently being released against the soybean aphid, Aphis glycines, in North American soybean fields. We conducted a
Giuseppina Andreotti et al.
Journal of biotechnology, 122(2), 274-284 (2005-11-18)
The purification and characterisation of the alpha-glucosidase from the marine mollusc Aplysia fasciata are reported. Overall substrate specificity of the pure enzyme for both hydrolytic and transglycosylation reactions was studied. Remarkable characteristics of this enzyme are indicated by the results
Roberto Consonni et al.
Journal of agricultural and food chemistry, 60(18), 4526-4534 (2012-04-19)
The saccharide profiles of 5 different botanical species in 86 Italian honey samples were investigated by ¹H and ¹H-¹³C NMR spectroscopy. Nineteen saccharides were identified in the aqueous extracts, namely, fructose, glucose, gentiobiose, isomaltose, kojibiose, maltose, maltulose, melibiose, nigerose, palatinose
J Peirce Beach et al.
Journal of chemical ecology, 29(5), 1203-1222 (2003-07-15)
The gustatory response of female Anaphes iole wasps to naturally occurring carbohydrates, a commercial food source, and host (Lygus lineolaris) frass was determined. Wasps responded to all 14 of the sugars at the highest concentration tested (2 M). At this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service