Skip to Content
Merck
All Photos(6)

Key Documents

T9531

Sigma-Aldrich

D-(+)-Trehalose dihydrate

≥99% (HPLC), from Saccharomyces cerevisiae

Synonym(s):

Trehalose dihydrate, α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · 2H2O
CAS Number:
Molecular Weight:
378.33
Beilstein:
5322018
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

Saccharomyces cerevisiae

Quality Level

Assay

≥99% (HPLC)

form

powder

optical activity

[α]20/D 174 to 182 °, c = 7% (w/v) in water

technique(s)

HPLC: suitable
PCR: suitable
cell based assay: suitable
cryopreservation: suitable

impurities

≤5.0% EtOH
<20 ppm Trace metals

color

white

mp

97-99  °C

solubility

water: 50 mg/mL, clear, colorless

application(s)

advanced drug delivery
agriculture
cell analysis
life science and biopharma
sample preparation

storage temp.

room temp

SMILES string

[H]O[H].[H]O[H].OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11.2H2O/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12;;/h3-20H,1-2H2;2*1H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-;;/m1../s1

InChI key

DPVHGFAJLZWDOC-PVXXTIHASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trehalose, also known as mycose or tremalose, is a nonreducing disaccharide of glucose commonly found in fungi, bacteria, and invertebrates. Its presence in these organisms highlights its significance in various biological processes and potential applications in biochemical research and cell culture studies.

Application

D-(+)-Trehalose dihydrate has been used:

  • as a transcription factor EB (TFEB) to study its effects on an autophagy-lysosomal system in human silicosis alveolar macrophages
  • as stabilizing excipient in spray-dried protein formulations
  • as a cryoprotectant to study its effects on mouse testicular cell suspensions

Biochem/physiol Actions

D-(+)-Trehalose shows anti-desiccant and cryopreservative activities. It acts as an osmolyte, and stress protectant and helps in the storage and transport of carbon. Trehalose is involved in the food, cosmetic, and pharmaceutical industries. It plays a vital role in the survival of certain insects and plants known as anhydrobionts during harsh environments such as high rates of dehydration.

Quality

Reduced metal ion content.

Preparation Note

Prepared by ion exchange chromatography.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental and theoretical electron density distribution of alpha,alpha-trehalose dihydrate
Stevens E D, et al.
Carbohydrate Research, 345(10), 1469-1481 (2010)
Trehalose production: exploiting novel approaches
Schiraldi C, et al.
Trends in Biotechnology, 20(10), 420- 425 (2002)
Shiyi Tan et al.
Life sciences, 257, 118043-118043 (2020-07-06)
Alveolar macrophages (AMs) are the primary targets of silicosis. Blockade of autophagy may aggravate the apoptosis of AMs. Trehalose (Tre), a transcription factor EB (TFEB) activator, may impact the autophagy-lysosomal system in AMs during silicosis. However, the mechanism by which
Trehalose: current use and future applications
Ohtake S and Wang Y J
Journal of Pharmaceutical Sciences, 100(6) , 2020-2053 (2011)
Trehalose metabolism in plants
Lunn J E, et al.
The Plant Journal, 79(4), 544-567 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service