Skip to Content
Merck
All Photos(1)

Documents

T2754

Sigma-Aldrich

D-(+)-Turanose

≥98%

Synonym(s):

3-O-α-D-Glucopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93771
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥98%

form

powder

optical activity

[α]20/D 74.8 to 76.8°, c = 4% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@@H]2[C@@H](O)[C@@H](O)COC2(O)CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-5-7(17)8(18)9(19)11(22-5)23-10-6(16)4(15)2-21-12(10,20)3-14/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12?/m1/s1

InChI key

SEWFWJUQVJHATO-PUVWEJBASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Turanose is an analog of sucrose that is not metabolized by higher plants but is used as a carbon source by many bacteria, fungi, and other organisms. It is taken up into plant cells by sucrose transporters and is involved in intracellular sugar signaling.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Study on alpha-glucosidases in four human colon malignant tumors developed into nude mice.
C Castilla et al.
The International journal of biochemistry, 13(3), 381-387 (1981-01-01)
G Potocki de Montalk et al.
FEBS letters, 471(2-3), 219-223 (2000-04-18)
Amylosucrase is a glucosyltransferase that synthesises an insoluble alpha-glucan from sucrose. The catalytic properties of the highly purified amylosucrase from Neisseria polysaccharea were characterised. Contrary to previously published results, it was demonstrated that in the presence of sucrose alone, several
J Defaye et al.
Carbohydrate research, 251, 1-15 (1994-01-03)
Dispirodioxanyl pseudotetrasaccharides 6-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 6-O-alpha-D-glucopyranosyl-beta-D-fructofuranose 1,2':2,1'-dianhydride, 5-O-alpha-D-glucopyranosyl-alpha-D-fructopyranose 5-O-alpha-D-glucopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, 4-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 4-O-alpha-D-glucopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, 4-O-beta-D-galactopyranosyl-alpha-D-fructofuranose 4-O-beta-D-galactopyranosyl-beta-D-fructopyranose 1,2':2,1'-dianhydride, and 3-O-alpha-D-glucopyranosyl-alpha-D-fructofuranose 3-O-alpha-D-glucopyranosyl-beta-D-fructofuranose 1,2':2,1'-dianhydride were respectively obtained, on a preparative scale, by dissolution of the isomeric glycosylfructoses palatinose, leucrose, maltulose, lactulose, and turanose in anhydrous hydrogen
Janet Hsu et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(5), 731-739 (2011-09-13)
Enzyme replacement therapies for lysosomal storage disorders are often hindered by suboptimal biodistribution of recombinant enzymes after systemic injection. This is the case for Pompe disease caused by acid α-glucosidase (GAA) deficiency, leading to excess glycogen storage throughout the body
J Defaye et al.
Carbohydrate research, 237, 223-247 (1992-12-31)
Selective activation of the ketose unit in the isomeric glycosylfructoses, palatinose, leucrose, maltulose, turanose and lactulose, with pyridinium poly(hydrogen fluoride) resulted in the almost quantitative formation of glycosylated difructose dianhydrides. The reaction preferentially involves a reactive fructofuranosyl oxocarbenium ion and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service