Skip to Content
Merck
All Photos(1)

Key Documents

E8640

Sigma-Aldrich

E-64d

powder, ≥98% (TLC)

Synonym(s):

(2S,3S)-trans-Epoxysuccinyl-L-leucylamido-3-methylbutane ethyl ester, EST

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30N2O5
CAS Number:
Molecular Weight:
342.43
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

E-64d, protease inhibitor

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

methanol: water (1:1): 5 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCOC(=O)[C@H]1O[C@@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCCC(C)C

InChI

1S/C17H30N2O5/c1-6-23-17(22)14-13(24-14)16(21)19-12(9-11(4)5)15(20)18-8-7-10(2)3/h10-14H,6-9H2,1-5H3,(H,18,20)(H,19,21)/t12-,13-,14-/m0/s1

InChI key

SRVFFFJZQVENJC-IHRRRGAJSA-N

Gene Information

human ... CAPN1(823)

Application

E-64d has been used as a lysosomal inhibitor in human acute promyelocytic leukemia NB4 cells[1] and Huh-7 cells.[2] It has also been used as a protease inhibitor in free calcium physiological tyrode solution for perfusion into the isolated heart samples.[3]

Biochem/physiol Actions

E-64d is an epoxysuccinyl peptide and an inhibitor of cysteine protease cathepsin B,[4] calpains 1 and 2.[5] E-64d by its cathepsin B protease inhibition functionality, may serve as a potential drug for treating traumatic brain injury (TBI)[5] and Alzheimer′s disease (AD).[6] It inhibits gametocyte surface antigen resulting in a decreased oocyst production in Plasmodium falciparum.[7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weidong Huang et al.
Scientific reports, 7(1), 13354-13354 (2017-10-19)
A cDNA library from a pool of all the seven silk glands from a tropical spider species was constructed. More than 1000 expressed sequence tag (EST) clones were created. Almost 65% of the EST clones were identified and around 50%
John P Lloyd et al.
Scientific reports, 9(1), 12122-12122 (2019-08-23)
Extensive transcriptional activity occurring in intergenic regions of genomes has raised the question whether intergenic transcription represents the activity of novel genes or noisy expression. To address this, we evaluated cross-species and post-duplication sequence and expression conservation of intergenic transcribed
Ethambutol neutralizes lysosomes and causes lysosomal zinc accumulation
Yamada D, et al.
Biochemical and Biophysical Research Communications, 471(1), 109-116 (2016)
E64d, a membrane-permeable cysteine protease inhibitor, attenuates the effects of parathyroid hormone on osteoblasts in vitro
Murray EJ, et al.
Metabolism and Disease, 46(9), 1090-1094 (1997)
Shingo Nakamura et al.
Frontiers in plant science, 8, 1840-1840 (2017-11-23)
Wild barley (

Questions

1–5 of 5 Questions  
  1. What can I dissolve Product E8640, E-64d, in?

    1 answer
    1. According to McGowan, E. B. et al., Biochem. Biophys. Res. Comm., 158(2), 432-435 (1989), this compound was dissolved in DMSO up to 200 mg/mL for use as an inhibitor of calpain in both intact and lysed platelets..

      Helpful?

  2. Where can solutions of Product E8640, E-64d, be stored?

    1 answer
    1. According to McGowan, E. B. et al., Biochem. Biophys. Res. Comm., 158(2), 432-435 (1989), solutions made in DMSO were stored at -70°C.

      Helpful?

  3. What is the difference between E-64, E-64c and E-64d?

    1 answer
    1. E-64c is a synthetic analog of E-64.  E-64d is the ethyl ester of E-64c.  E-64c is not cell permeable, but E-64d is cell permeable.  E-64d is hydrolyzed in cell culture to E-64c.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service