Skip to Content
Merck
All Photos(1)

Key Documents

77170

Sigma-Aldrich

Pepstatin A

≥100,000 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Beilstein:
2201362
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

Streptomyces sp.

Quality Level

form

powder

specific activity

≥100,000 U/mg

mp

233 °C (dec.) (lit.)

solubility

ethanol: soluble 1 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)[C@@H](O)CC(O)=O

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23-,24-,25-,26-,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-LXTPJMTPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pepstatin A is suitable for use:
  • in the inhibitor cocktail in fluorometric matrix metalloproteinase assay
  • as a protease inhibitor during homogenization of tissue
  • as a protease inhibitor during extraction of cells
  • to determine the molar concentrations of cathepsin D by titration with pepstatin A
  • in inhibiting cathepsin D
It can be used in conjunction with E64-d and Leupeptin A to inhibit the degradation of autophagic cargo inside autophagosomes. For this application, the working concentration is typically between 1-10 μM.

Biochem/physiol Actions

Pepstatin A is an inhibitor of acid proteases (aspartyl peptidases). It forms a 1:1 complex with proteases such as pepsin, renin, cathepsin D, bovine chymosin, and protease B (Aspergillus niger). The inhibitor is highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases. Solubilized γ-secretase and retroviral protease are also inhibited by Pepstatin A. It has been used to characterize proteases from several sources.
Potent inhibitor of acid proteases, including pepsin, renin and cathepsin D and many microbial aspartic proteases. Effective concentration 1 μM. Stable for about 1 day.

Unit Definition

1 U corresponds to the amount of inhibitor which reduces the activity of pepsin by 1 U (1 U corresponds to the amount of enzyme which increases the absorbance at 280 nm by 0.001 per minute at pH 2.0 and 37°C (Hemoglobin, Cat. No. 51290, as substrate, in an endvolume of 16 ml)); 10′000 absorbance-U as described above are equivalent to ∼1 Bergmeyer-U (when 1 pepsin-U is the amount of enzyme which hydrolyzes 1 μmol acetyl-L-phenylalanyl-3,5-diiodo-L-tyrosin per minute at pH 2.0 and 37°C)

Preparation Note

Pepstatin can be dissolved at 1 mg/ml in 10% (v/v) acetic acid in methanol (9:1 methanol:acetic acid). The inclusion of acetic acid may be necessary to dissolve this peptide in methanol or DMSO. It has been dissolved at 1 to 2 mg/ml in ethanol, but heat may be required for complete dissolution. Solutions of Pepstatin A can be heated as high as 60 °C without any decomposition of the peptide.
Stock solutions at 1 mg/ml are stable at least a week at 4 °C. A 1 mM solution in methanol or DMSO is stable for months at -20 °C. If solutions become darker yellow, the reagent is hydrolyzing.
A working concentration of 1 μM is stable for at least one day at room temperature.

Other Notes

Strong inhibitor of acid proteases - pepsin, cathepsin D, renin

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ferreras et al.
FEBS letters, 486(3), 247-251 (2000-12-20)
The angiogenesis inhibitor endostatin is a fragment of the NC1 domain of collagen XVIII. The generation of endostatin has been investigated only in murine hemangioendothelioma cell cultures and was ascribed to cathepsin L. Distinct endostatin-like fragments were detected in human
Kavindra Kumar Kesari et al.
International journal of radiation biology, 86(4), 334-343 (2010-04-01)
To investigate the effect of 2.45 GHz microwave radiation on rat brain of male wistar strain. Male rats of wistar strain (35 days old with 130 +/- 10 g body weight) were selected for this study. Animals were divided into
Protease inhibitors produced by microorganisms.
H Umezawa
Acta biologica et medica Germanica, 36(11-12), 1899-1915 (1977-01-01)
F G Giancotti et al.
The Journal of cell biology, 118(4), 951-959 (1992-08-01)
The alpha 6 beta 4 integrin is a receptor involved in the interaction of epithelial cells with basement membranes. This integrin is unique among the known integrins in that its beta 4 subunit has a large cytoplasmic domain. The function
Anne Kunath et al.
Biomedicines, 9(2) (2021-02-13)
Increased plasma and adipose tissue protease activity is observed in patients with type 2 diabetes and obesity. It has been proposed that specific proteases contribute to the link between obesity, adipose tissue inflammation and metabolic diseases. We have recently shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service