Skip to Content
Merck
All Photos(1)

Key Documents

C5732

Sigma-Aldrich

CA-074

≥99% (TLC)

Synonym(s):

(L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H29N3O6
CAS Number:
Molecular Weight:
383.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥99% (TLC)

form

powder

solubility

methanol: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCCNC(=O)[C@H]1O[C@@H]1C(=O)N[C@@H]([C@H](C)CC)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C18H29N3O6/c1-4-8-19-15(22)13-14(27-13)16(23)20-12(10(3)5-2)17(24)21-9-6-7-11(21)18(25)26/h10-14H,4-9H2,1-3H3,(H,19,22)(H,20,23)(H,25,26)/t10?,11-,12-,13-,14-/m0/s1

InChI key

ZEZGJKSEBRELAS-NSIINPIOSA-N

Application

CA-074 has been used as a cathepsin B inhibitor:
  • to study its effect on pseudotyped viruses bearing S proteins harboring R642M and N714K using HeLa cells
  • to determine the role of autophagy in glucose deprivation (GD)-induced damage
  • to study its effects on neuronal function upon exposure to His-cathepsin B (CATB)

Biochem/physiol Actions

CA-074 is a potent and a selective inhibitor of cathepsin B. It is useful to study the biological function of cathepsin B in vivo. Studies show that CA-074 can also inhibit cathepsin L under reducing conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Nicastro et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(30), 10493-10498 (2005-07-16)
The Josephin domain plays an important role in the cellular functions of ataxin-3, the protein responsible for the neurodegenerative Machado-Joseph disease. We have determined the solution structure of Josephin and shown that it belongs to the family of papain-like cysteine
Masashi Asai et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(10), 3720-3730 (2011-07-13)
γ-Secretase catalyzes the cleavage of the intramembrane region of the Alzheimer amyloid precursor protein (APP), generating p3, amyloid-β peptide (Aβ), and the APP intracellular domain (AICD). Although a γ-secretase inhibitor has been shown to cause an accumulation of the APP
Kazuya Shirato et al.
Journal of virology, 91(1) (2016-11-01)
Human coronavirus 229E (HCoV-229E), a causative agent of the common cold, enters host cells via two distinct pathways: one is mediated by cell surface proteases, particularly transmembrane protease serine 2 (TMPRSS2), and the other by endosomal cathepsin L. Thus, specific
Clément Naudin et al.
PloS one, 6(9), e25577-e25577 (2011-10-08)
Cysteine cathepsins have emerged as new players in inflammatory lung disorders. Their activities are dramatically increased in the sputum of cystic fibrosis (CF) patients, suggesting that they are involved in the pathophysiology of CF. We have characterized the cathepsins in
M Murata et al.
FEBS letters, 280(2), 307-310 (1991-03-25)
A series of new epoxysuccinyl peptides were designed and synthesized to develop a specific inhibitor of cathepsin B. Of these compounds, N-(L-3-trans-ethoxycarbonyloxirane-2-carbonyl)-L-isoleucyl-L-proli ne (compound CA-030) and N-(L-3-trans-propylcarbamoyloxirane-2-carbonyl)-L-isoleucyl-L-prol ine (compound CA-074) were the most potent and specific inhibitors of cathepsin B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service