Skip to Content
Merck
All Photos(4)

Documents

E3132

Sigma-Aldrich

E-64

protease inhibitor

Synonym(s):

trans-Epoxysuccinyl-L-leucylamido(4-guanidino)butane, L-trans-3-Carboxyoxiran-2-carbonyl-L-leucylagmatine, N-(trans-Epoxysuccinyl)-L-leucine 4-guanidinobutylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27N5O5
CAS Number:
Molecular Weight:
357.41
Beilstein:
1405664
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

powder

solubility

water: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCNC(N)=N

InChI

1S/C15H27N5O5/c1-8(2)7-9(20-13(22)10-11(25-10)14(23)24)12(21)18-5-3-4-6-19-15(16)17/h8-11H,3-7H2,1-2H3,(H,18,21)(H,20,22)(H,23,24)(H4,16,17,19)/t9-,10+,11+/m0/s1

InChI key

LTLYEAJONXGNFG-HBNTYKKESA-N

Looking for similar products? Visit Product Comparison Guide

General description

E-64 is a cysteine protease inhibitor that was isolated from the mold Aspergillus japonicus TPR-64. E-64 is also known as N-[N-(L-3-trans-carboxyoxiran-2-carbonyl)-L-leucyl]-agmatine. E-64 effectively inhibits various cysteine proteases, in particular:
  • cathepsin K
  • cathepsin L
  • cathepsin S
E-64 also acts against other enzymes, such as:
  • calpain
  • cathepsin B
  • cathepsin H
  • papain

Application

E-64 is an effective ligand for affinity purification of cysteine proteases. When coupled to a thiolated affinity matrix, binding is no longer irreversible, but specificity is retained.

Biochem/physiol Actions

E-64 is an irreversible, potent, and highly selective cysteine protease inhibitor. E-64 does not react with the functional thiol group of non-protease enzymes, such as L-lactate dehydrogenase or creatine kinase. E-64 will not inhibit serine proteases (except trypsin) like other cysteine protease inhibitors, leupeptin and antipain. The trans-epoxysuccinyl group (active moiety) of E-64 irreversibly binds to an active thiol group in many cysteine proteases, such as papain, actinidase, and cathepsins B, H, and L to form a thioether linkage. E-64 is a very useful cysteine protease inhibitor for use in in vivo studies because it has a specific inhibition, it is permeable in cells and tissues and has low toxicity.

Preparation Note

E-64 is soluble in water. A 20 mg/ml solution can be prepared in water (heat may be needed). A suggested stock solution is a 1 mM aqueous solution. The effective concentration for use as a protease inhibitor is 1 to10 μM. Aqueous stock solutions are stable for months at -20 °C. Diluted solutions are stable for days at neutral pH. E-64 is stable from pH 2-10, but is unstable in ammonia or in HCl. E-64 is also soluble in DMSO, a 10 mM solution can be prepared in dry DMSO and stored at -20 °C. Subsequent dilutions were in culture medium. Solutions for injection were prepared by dissolving E-64 in 0.9% sodium chloride or in a minimum amount of saturated sodium bicarbonate followed by dilution with 0.9% sodium chloride (after adjusting the pH to 7.0 with acetic acid).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Gobbetti et al.
The American journal of pathology, 180(1), 141-152 (2011-11-10)
Proteases and proteinase-activated receptor (PAR) activation are involved in several intestinal inflammatory conditions. We hypothesized that serine proteases and PAR activation could also modulate the intestinal injury induced by ischemia-reperfusion (I-R). C57Bl/6 mice were subjected to 90 minutes of intestinal
F D Nascimento et al.
Journal of dental research, 90(4), 506-511 (2011-01-21)
Matrix metalloproteinases (MMPs) are important in dentinal caries, and analysis of recent data demonstrates the presence of other collagen-degrading enzymes, cysteine cathepsins, in human dentin. This study aimed to examine the presence, source, and activity of cysteine cathepsins in human
L Latonen et al.
Oncogene, 30(7), 790-805 (2010-10-20)
The ubiquitin-proteasome pathway is essential for most cellular processes, including protein quality control, cell cycle, transcription, signaling, protein transport, DNA repair and stress responses. Hampered proteasome activity leads to the accumulation of polyubiquitylated proteins, endoplastic reticulum (ER) stress and even
P M C Scaffa et al.
Journal of dental research, 91(4), 420-425 (2012-01-24)
The co-expression of MMPs and cysteine cathepsins in the human dentin-pulp complex indicates that both classes of enzymes can contribute to the endogenous proteolytic activity of dentin. Chlorhexidine (CHX) is an efficient inhibitor of MMP activity. This study investigated whether
Yohei Osako et al.
The FEBS journal, 277(21), 4412-4426 (2010-09-21)
Calpain 7, a mammalian ortholog of yeast Cpl1/Rim13 and fungal PalB, is an atypical calpain that lacks a penta-EF-hand domain. Previously, we reported that a region containing a tandem repeat of microtubule-interacting and transport (MIT) domains in calpain 7 interacts

Articles

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service