Skip to Content
Merck
All Photos(1)

Key Documents

T2376

Sigma-Aldrich

L-Thyroxine

≥98% (HPLC)

Synonym(s):

3,3′,5,5″-Tetraiodo-L-thyronine, 3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(I)2OC6H2(I)2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
776.87
Beilstein:
2228515
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic
form:
powder

biological source

synthetic

sterility

non-sterile

Assay

≥98% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

223 °C (dec.) (lit.)

solubility

4 M NH4OH in methanol: 50 mg/mL, clear to hazy, yellow

shipped in

ambient

storage temp.

room temp

SMILES string

N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H11I4NO4/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23/h1-2,4-5,12,21H,3,20H2,(H,22,23)/t12-/m0/s1

InChI key

XUIIKFGFIJCVMT-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

L-Thyroxine was used to study the light-induced hormone conversion of T4 to T3 in regulation of photoperiodic response of gonads in Japanese quail. The product was used as a test compound for studying the impact of numerous hormones on the surface morphology of rat testicular cells in culture.

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and, thus, protein synthesis.
L-Thyroxine is an iodine containing hormone produced from thyroglobulin in the thyroid follicular cells. L-Thyroxine reduces total cholesterol level, low-density lipoprotein cholesterol as well as waist to hip ratio in subclinical hypothyroidism pateints. Hence, facilitates in improving cardiovascular risk factor as well as can enhance symptoms of tiredness.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

L-Thyroxine dissolves in 4M ammonium hydroxide in Methanol at 50 mg/ml to yield a clear to hazy yellow to dark yellow solution.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J C Hutson
The American journal of anatomy, 151(1), 55-69 (1978-01-01)
The purpose of this investigation was to study the effects of various hormones on the surface morphology of 20-day-old rat testicular cells in culture. Aggregates containing primarily Sertoli cells and germinal cells were obtained by enzymatic digestion. The surface morphology
Tom Sarraude et al.
PeerJ, 8, e10175-e10175 (2020-10-23)
Maternal thyroid hormones (THs) are known to be crucial in embryonic development in humans, but their influence on other, especially wild, animals remains poorly understood. So far, the studies that experimentally investigated the consequences of maternal THs focused on short-term
Turan Demircan et al.
Data in brief, 29, 105179-105179 (2020-02-15)
The presented data article reports protein expression profiles during a time course of limb regeneration in the highly regenerative neotenic and regeneration-deficient metamorphic axolotl (Ambystoma mexicanum). A protein database was first generated from transcriptome data, which was used concomitantly with
Takashi Yoshimura et al.
Nature, 426(6963), 178-181 (2003-11-14)
Reproduction of many temperate zone birds is under photoperiodic control. The Japanese quail is an excellent model for studying the mechanism of photoperiodic time measurement because of its distinct and marked response to changing photoperiods. Studies on this animal have
Inés Martín-Lacave et al.
Journal of anatomy, 214(3), 301-309 (2009-02-28)
C cells are primarily known for producing calcitonin, a hypocalcemic and hypophosphatemic hormone. Nevertheless, besides their role in calcium homeostasis, C cells may be involved in the intrathyroidal regulation of follicular cells, suggesting a possible interrelationship between the two endocrine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service