A9037
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in goat
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti Rat, Anti Rat Hrp
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
goat
conjugate
peroxidase conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct ELISA: 1:10,000
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Related Categories
General description
IgGs are glycoprotein antibodies that modulate several immune responses. Rat IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rat IgG conjugated to a detectable substrate is a useful tool for the analysis of target proteins. Anti-Rat IgG (whole molecule)-Peroxidase antibody binds all rat immunoglobulins.
IgG (immunoglobulin γ) is a monomeric antibody containing a γ Fc region as a part of heavy chain, which is linked to the Fab arm through a hinge region. It is the most abundant serum antibody and constitutes to 70% of the total immunoglobulins.
IgG (immunoglobulin γ) is a monomeric antibody containing a γ Fc region as a part of heavy chain, which is linked to the Fab arm through a hinge region. It is the most abundant serum antibody and constitutes to 70% of the total immunoglobulins.
Immunogen
Purified rat IgG
Application
Anti-Rat IgG (whole molecule)-Peroxidase antibody is suitable for use in ELISA and immunoblot at 1:5000 dilutions.
Lysates generated from primary cells isolated from lymphoid organs of P. monodon shrimp were analyzed by western blot using HRP-conjugated goat anti-rat as the secondary antibody at a 1:4000 dilution.
Biochem/physiol Actions
IgG (immunoglobulin γ), upon epitope binding, mediates the activation of classical complement pathway. Interaction between the host cell membrane and IgG antibody results in type II hypersensitivity reaction. Decreased serum levels of IgG contributes to common variable immunodeficiency, thus making the individual easily prone to infections.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative
Preparation Note
Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Trevor R F Smith et al.
Vaccine, 35(21), 2840-2847 (2017-04-18)
Respiratory syncytial virus (RSV) is a massive medical burden in infants, children and the elderly worldwide, and an effective, safe RSV vaccine remains an unmet need. Here we assess a novel vaccination strategy based on the intradermal delivery of a
T Orth et al.
European journal of clinical investigation, 29(11), 929-939 (1999-12-03)
In sera from patients with autoimmune liver diseases, e. g. primary sclerosing cholangitis (PSC) and autoimmune hepatitis, anti-neutrophil cytoplasmic antibodies (ANCAs) can be found. Until now, no animal model of ANCA induction in liver disease has been described. In this
Wanchai Assavalapsakul et al.
Journal of virology, 80(1), 262-269 (2005-12-15)
The yellow head virus is a positive-sense, single-stranded RNA virus that causes significant mortality in farmed penaeid shrimp. This study sought to isolate and characterize the receptor protein used by the virus to gain entry into Penaeus monodon Oka (lymphoid)
Máximo Lopez Medus et al.
Scientific reports, 7(1), 8788-8788 (2017-08-20)
Nearly one third of the eukaryotic proteome traverses the secretory pathway and most of these proteins are N-glycosylated in the lumen of the endoplasmic reticulum. N-glycans fulfill multiple structural and biological functions, and are crucial for productive folding of many
Stéphane Mesnage et al.
Nature communications, 5, 4269-4269 (2014-07-01)
Carbohydrate recognition is essential for growth, cell adhesion and signalling in all living organisms. A highly conserved carbohydrate binding module, LysM, is found in proteins from viruses, bacteria, fungi, plants and mammals. LysM modules recognize polysaccharides containing N-acetylglucosamine (GlcNAc) residues
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service