Skip to Content
Merck
All Photos(1)

Documents

AP136P

Sigma-Aldrich

Goat Anti-Rat IgG Antibody, HRP conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Goat anti-rat IgG affinity purified antibody, HRP conjugated.

Specificity

Specific for rat IgG, heavy and light chain.

Application

Detect Rat IgG using this Goat anti-Rat IgG Antibody, HRP conjugate validated for use in ELISA, IH & WB.
Immunohistochemistry:
1:500-1:5,000 dilution can be used.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated on Enzyme-linked Immunosorbent Assay.

ELISA and Western Blots:
1:5,000-1:100,000 dilution can be used..

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 7.6 with 15 mg/mL BSA.
Reconstitute with 2 mL of sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months.
After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.
WARNING: Use of sodium azide as a preservative will substantially inhibit the enzyme activity of HRP.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Counteraction of tetherin antiviral activity by two closely related SIVs differing by the presence of a Vpu gene.
Nikovics, K; Dazza, MC; Ekwalanga, M; Mammano, F; Clavel, F; Saragosti, S
Testing null
Mark A Csandl et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(6), 857-866 (2016-04-14)
Active efflux of both drugs and organic anion metabolites is mediated by the multidrug resistance proteins (MRPs). MRP1 (ABCC1), MRP2 (ABCC2), MRP3 (ABCC3), and MRP4 (ABCC4) have partially overlapping substrate specificities and all transport 17β-estradiol 17-(β-d-glucuronide) (E217βG). The cysteinyl leukotriene
Felix Wiedmann et al.
Molecular biology of the cell, 30(12), 1425-1436 (2019-04-11)
Two pore-domain potassium (K2P) channels mediate potassium background currents that stabilize the resting membrane potential and facilitate action potential repolarization. In the human heart, hK2P17.1 channels are predominantly expressed in the atria and Purkinje cells. Reduced atrial hK2P17.1 protein levels
Quan Chen et al.
Molecular medicine reports, 14(1), 202-208 (2016-05-14)
Ultraviolet (UV) radiation-induced immunosuppression leading to skin cancer has received increased attention in previous years. The present study aimed to investigate the immunoprotection offered by Anthelios sunscreen in a mouse model of Candida albicans‑induced delayed‑type hypersensitivity. Anthelios sunscreen was applied
N V Andreeva et al.
Cell stress & chaperones, 21(4), 727-733 (2016-04-20)
Heat shock proteins including the major stress protein HSP70 support intracellular homeostasis and prevent protein damage after a temperature increase and other stressful environmental stimuli, as well as during aging. We have shown earlier that prolonged administration of recombinant human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service