Skip to Content
Merck
All Photos(1)

Key Documents

F6258

Sigma-Aldrich

Anti-Rat IgG (whole molecule)–FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rat IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:32

storage temp.

2-8°C

target post-translational modification

unmodified

General description

An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Rat has five immunoglobulin classes- IgM, IgG, IgA, IgD and IgGE. IgG has four subclasses- IgG1, IgG2a, IgG2b and IgG2c. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Rat IgG (whole molecule)-FITC antibody is specific for all rat IgG subclasses. Goat anti-rat IgG is purified by affinity isolation and then conjugated to FITC.
IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. It regulates several functions such as complement activation and phagocytosis and hence plays a pivotal role in facilitating cytological immune responses. Anti-rat IgG (whole molecule)-FITC antibody can be used as a secondary antibody in indirect immunofluorescence. Goat anti- rat IgG (whole molecule)-FITC antibody reacts specifically with all rat immunoglobulins.

Immunogen

Purified rat IgG
Purified rat IgG.

Application

Anti-Rat IgG (whole molecule)−FITC antibody has been used in immunofluorescence measurement of antibody-treated cells and staining of monolayers.
Anti-Rat IgG (whole molecule)-FITC antibody may be used for immunofluorescent labeling of rat spleen cells. For immunofluorescence of rat cartilage tissue sections, a concentration of 5-10 μg/ml was used. The range of dilutions of anti-rat IgG used for immunofluorescence is between 1:10 and 1:250 in various studies.. The antibody was also used in flow cytometry applications.
Anti-rat IgG (whole molecule)-FITC antibody (diluted 1:20 in PBS) can be used as a secondary antibody in immunolabelling. It can also be used in flow cytometry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Cassis et al.
JCI insight, 3(15) (2018-08-10)
Sodium-glucose cotransporter 2 (SGLT2) inhibitors have pleiotropic properties beyond blood glucose-lowering effects and modify important nonglycemic pathways, leading to end-organ protection. SGLT2 inhibitors display renoprotective effects in diabetic kidney disease, which creates a rationale for testing the therapeutic potential of
Yung-I Lee et al.
American journal of botany, 100(11), 2121-2131 (2013-10-19)
Protocorm-like bodies (PLBs) of orchids are important in orchid micropropagation and outwardly resemble somatic embryos in form and development. To determine whether PLBs are truly embryogenetic, we compared PLBs with somatic embryos and zygotic embryos to determine whether they had
Receptors involved in lymphocyte homing: relationship between a carbohydrate-binding receptor and the MEL-14 antigen.
Yednock T A, et al.
The Journal of cell biology, 104(3), 725-731 (1987)
Aged HCT-8 cell monolayers support Cryptosporidium parvum infection
Sifuentes L Y, et al.
Applied and Environmental Microbiology, 73(23), 7548-7551 (2007)
M Niimi et al.
Journal of immunology (Baltimore, Md. : 1950), 161(10), 5331-5337 (1998-11-20)
Resting B (rB) cells are known to be incompetent APCs in vitro, which alone can induce specific unresponsiveness to single minor histocompatibility (miH) Ags and, when combined with CD40 pathway blockade, can induce hyporesponsiveness to MHC molecules in vivo. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service