Skip to Content
Merck
All Photos(1)

Key Documents

A6154

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Rabbit IgG (whole molecule)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat Anti-Rabbit IgG (whole molecule)-Peroxidase antibody is immunospecific for rabbit IgG by immunoelectrophoresis against normal serum and rabbit IgG, prior to conjugation.

Immunogen

purified rabbit IgG

Application

C33A cell lysates were analyzed by western blot using HRP-conjugated goat anti-rabbit IgG as the secondary antibody. Protein aliquots from colonic mucosal extracts (μg) were anlayzed by western blot using HRP-conjugated goat anti-rabbit IgG as the secondary antibody at a 1:5000 dilution in TBSt for 1 hr at room temperature.
Co-immunoprecipation and western blot analysis of C33A cell lysates were performed using HRP conjugated goat anti-rabbit IgG as the secondary antibody.
Goat Anti-Rabbit IgG (whole molecule)-Peroxidase antibody has been used for western blot (1:6,000), and ELISA assays.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan de Vogel et al.
Carcinogenesis, 29(2), 398-403 (2008-01-05)
Epidemiological and animal model studies suggest that a high intake of heme, present in red meat, is associated with an increased risk of colon cancer. The aim of this study was to elucidate the effects of dietary heme on colonic
Patrick Eldin et al.
Nucleic acids research, 42(3), 1698-1710 (2013-11-02)
The human immunodeficiency virus type 1 (HIV-1) Vpr protein binds to the cellular uracil-DNA glycosylase UNG2 and induces its degradation through the assembly with the DDB1-CUL4 ubiquitin ligase complex. This interaction counteracts the antiviral activity exerted by UNG2 on HIV-1
Sandra V Ashton et al.
Arteriosclerosis, thrombosis, and vascular biology, 25(1), 102-108 (2004-10-23)
Invasion of uterine spiral arteries by extravillous trophoblasts in the first trimester of pregnancy results in loss of endothelial and musculoelastic layers. This remodeling is crucial for an adequate blood supply to the fetus with a failure to remodel implicated
Pascal Nurwakagari et al.
Journal of molecular endocrinology, 38(1-2), 259-275 (2007-02-13)
Glycoprotein hormone receptors such as the lutropin/chorionic gonadotropin receptor (LHR) are characterized by a large N-terminal ectodomain (ECD), which is responsible for hormone-receptor interactions. For the closely related TSH receptor (TSHR), it has been proposed that the ECD also serves
Steffen Erkelenz et al.
Retrovirology, 12, 29-29 (2015-04-19)
The viral regulatory protein Tat is essential for establishing a productive transcription from the 5'-LTR promoter during the early phase of viral gene expression. Formation of the Tat-encoding mRNAs requires splicing at the viral 3'ss A3, which has previously been

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service