Skip to Content
Merck
All Photos(2)

Key Documents

A9169

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti Rabbit Antibody - Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, Anti Rabbit Antibody Sigma

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

technique(s)

direct ELISA: 1:50,000 using using 5 μg/ml of rabbit IgG for coating and OPD substrate
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400
western blot (chemiluminescent): 1:80,000-1:160,000 using using β-actin in total cell extract of HeLa cells (5-10 μg per well)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. IgG is a major class of immunoglobulin. Rabbit IgG is further divided into five classes- IgM, IgG, IgA, IgD and IgE. It has only one IgG variant with one interheavy disulfide bridge.
Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins.
Goat Anti-Rabbit IgG (whole molecule)-Peroxidase antibody binds all rabbit Igs.

Specificity

Specificity of the Peroxidase Conjugated Anti-Rabbit IgG antibodies is determined by immunoelectrophoresis (IEP) versus normal rabbit serum and rabbit IgG.

Immunogen

purified rabbit IgG

Application

Anti-Rabbit IgG (whole molecule)–Peroxidase antibody has been used in western blotting.
Goat Anti-Rabbit IgG (whole molecule)-Peroxidase antibody can be used for immunoblot assays at 1:1,000. The antibody can also be used for ELISA (1:5,000) and IHC (1:400) applications.
Cell-associated protein samples from E. coli cultures were analyzed by western blot using HRP-conjugated goat anti-rabbit IgG as the secondary antibody.
Co-immunoprecipation and western blot analysis of C33A cell lysates were performed using HRP conjugated goat anti-rabbit IgG as the secondary antibody.

Biochem/physiol Actions

Immunoglobulins are glycoprotein antibodies that modulate several immune responses. Immunoglobulin G participates in hypersensitivity type II and type III. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by two-step glutaraldehyde method.

Storage and Stability

For continuous use, store at 0-5EC. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MDR1-deficient genotype in Collie dogs hypersensitive to the P-glycoprotein substrate ivermectin.
Roulet A, et al.
European Journal of Pharmacology, 460(2-3), 85-91 (2003)
L J M Carvalho et al.
Scandinavian journal of immunology, 59(4), 363-372 (2004-03-31)
The immunogenicity and protective efficacy of various antigen-adjuvant formulations derived either from the merozoite-surface protein-3 (MSP-3) or the glutamate-rich protein (GLURP) of Plasmodium falciparum were evaluated in Saimiri sciureus monkeys. These proteins were selected for immunogenicity studies based primarily on
Deficits in GABAB receptor system in schizophrenia and mood disorders: a postmortem study.
Fatemi SH, et al.
Schizophrenia Research, 128(1-3), 37-43 (2011)
Expression of GABA A α2-, β1-and γ-receptors are altered significantly in the lateral cerebellum of subjects with schizophrenia, major depression and bipolar disorder.
Fatemi SH, et al.
Translational Psychiatry, 3(9), e303-e303 (2013)
Sarah J Coulthurst et al.
The Journal of biological chemistry, 283(35), 23739-23753 (2008-06-20)
Erwinia carotovora subsp. atroseptica is an enterobacterial phytopathogen causing economically significant soft rot disease. Pathogenesis is mediated by multiple secreted virulence factors, many of which are secreted by the type II (Out) secretion system. DsbA catalyzes the introduction of disulfide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service