Skip to Content
Merck
All Photos(3)

Key Documents

A5754

Sigma-Aldrich

3-Amino-9-ethylcarbazole

≥95% (HPLC), powder

Synonym(s):

9-Ethylcarbazol-3-amine, AEC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2
CAS Number:
Molecular Weight:
210.27
Beilstein:
174969
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder

Assay

≥95% (HPLC)

form

powder

impurities

≤3% solvent

solubility

ethanol: acetic acid (2:1): 50 mg/mL, clear to hazy

storage temp.

2-8°C

SMILES string

CCn1c2ccccc2c3cc(N)ccc13

InChI

1S/C14H14N2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2,15H2,1H3

InChI key

OXEUETBFKVCRNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Amino-9-ethylcarbazole serves as chromogen in immunocytochemistry and produces a red color.

Specificity

3-Amino-9-ethylcarbazole is a peroxidase substrate suitable for use in immunoblotting and immunohistochemical staining procedures. This substrate produces an insoluble end product that is red in color and can be observed visually.

Application

3-Amino-9-ethylcarbazole has been used to visualize the bound primary antibody by using a streptavidin method. It has also been used in visualizing tissue sections from tumour samples.

Substrates

Substrate for peroxidase.

Analysis Note

Actual content given on label

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patterns of angiogenesis in nonsmall-cell lung carcinoma
Offersen BV, et al.
Cancer, 91(8), 1500-1509 (2001)
VEGF-C and COX-2 expression in papillary thyroid cancer
Siironen P, et al.
Endocrine-Related Cancer, 13(2), 465-473 (2006)
Cristan Herbert et al.
Respirology (Carlton, Vic.), 20(8), 1206-1212 (2015-08-21)
Exacerbations of allergic asthma are often triggered by respiratory viral infections. We have previously shown that in a T-helper type 2 (Th2)-biased cytokine environment, mouse and human airway epithelial cells (AEC) exhibit increased expression of pro-inflammatory and anti-viral genes in
Rihab Bouchareb et al.
Circulation, 132(8), 677-690 (2015-08-01)
Mendelian randomization studies have highlighted that lipoprotein(a) [Lp(a)] was associated with calcific aortic valve disease (CAVD). Lp(a) transports oxidized phospholipids with a high content in lysophosphatidylcholine. Autotaxin (ATX) transforms lysophosphatidylcholine into lysophosphatidic acid. We hypothesized that ATX-lysophosphatidic acid could promote
Viktor H Koelzer et al.
Translational research : the journal of laboratory and clinical medicine, 166(2), 207-217 (2015-03-24)
Tumor budding (single tumor cells or small tumor cell clusters) at the invasion front of colorectal cancer (CRC) is an adverse prognostic indicator linked to epithelial-mesenchymal transition. This study characterized the immunogenicity of tumor buds by analyzing the expression of

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service