Skip to Content
Merck
All Photos(1)

Documents

AP112

Sigma-Aldrich

Goat Anti-Human IgG Antibody

2 mg/mL, Chemicon®

Synonym(s):

Anti-Human IgG antibody, Goat anti-Human IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Reacts with Human IgG (H&L), as well as the light chains from all human immunoglobulin classes. Monospecific on IEP against whole human serum. Single band on IEP at a concentration of 10 mg/mL when run against rabbit anti goat whole serum

Immunogen

Highly purified Human IgG isolated from pooled normal human serum.

Application

Goat anti-Human IgG Antibody is an antibody against Human IgG for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Provided at 100 μg/100 μL in PBS, pH 7.4, containing 0.2% sodium merthiolate as a preservative.

Storage and Stability

Store at +2-8°C for up to 6 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiachong Shi et al.
Bioengineered, 13(3), 7564-7578 (2022-03-15)
Study has suggested that long non-coding RNA DOCK9 antisense RNA2 (LncRNA DOCK9-AS2) may play an important role in atherosclerosis, but the specific role is unclear. In this article, we aim to explore the role and mechanism of DOCK9-AS2 in the
A new nanoconstruct for epidermal growth factor receptor-targeted photo-immunotherapy of ovarian cancer.
Mir, Y; Elrington, SA; Hasan, T
Nanomedicine: Nanotechnology, Biology, and Medicine null
Shasha Ren et al.
Journal of experimental & clinical cancer research : CR, 38(1), 388-388 (2019-09-07)
Hypoxia is a key feature of breast cancer, which affects cancer development, metastasis and metabolism. Previous studies suggested that circular RNAs (circRNAs) could participate in cancer progression and hypoxia regulation. This study aimed to investigate the role of circRNA differentially
Yang Shen et al.
Experimental and therapeutic medicine, 22(2), 795-795 (2021-06-08)
MicroRNAs (miRs) have been implicated in the development of acute pancreatitis (AP). However, the role and potential mechanism of miR-9 in AP progression remains unclear. Caerulein-treated AR42J cells were used as a cellular model of AP. Results revealed caerulein triggered
Ling Tao et al.
BioMed research international, 2020, 7183629-7183629 (2020-11-03)
Long noncoding RNA cancer susceptibility candidate 2 (CASC2) has been reported to play an anticancer role in papillary thyroid cancer (PTC). Radioiodine (131I) is a common option for the treatment of PTC. However, the role and mechanism of CASC2 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service