Skip to Content
Merck
All Photos(2)

Key Documents

AEC101

Sigma-Aldrich

AEC Staining Kit

liquid

Synonym(s):

ACE substrate kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116134
NACRES:
NA.32

form

liquid

Quality Level

usage

sufficient for ≥1,000 tests (150 ml substrate solution)

storage temp.

2-8°C

Application

AEC Staining Kit has been used as a substrate
  • for developing the extravidin peroxidase antibody in cell-seeded matrice sections using histology and immunohistochemistry methods
  • for streptavidin-horseradish peroxidase in peripheral blood mononuclear cells in ex vivo enzyme-linked immunospot (ELISPOT) assay
  • for mouse IgG biotinylated secondary antibody in immunohistostaining of epididymal fat pads

Packaging

Kit contains 3 mL of concentrated acetate buffer, 3-amino-9-ethylcarbazole (AEC) chromogen and hydrogen peroxide in a dropper bottle for easy dispensing. Directions for use with each application are included.

Caution

Stable at least one year at 2-8 °C.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirti Kajal et al.
Scientific reports, 9(1), 4073-4073 (2019-03-13)
Vasculogenesis and angiogenesis are process of formation of blood vessels. Blood vessels are evolved to distribute nutrients and oxygen to distant organs. These vessels are crucial for growth and repair of wounded tissue. During tumor condition there occurs imbalance in
B Volck et al.
Osteoarthritis and cartilage, 9(3), 203-214 (2001-04-13)
The presence of YKL-40 (human cartilage glycoprotein 39) in synovium, cartilage and synovial fluid (SF) from knee joints of patients with rheumatoid arthritis and osteoarthritis (OA) were related to histopathological changes in synovium and cartilage and to serum YKL-40 and
Kaiyuan Ni et al.
Matter, 1(5), 1331-1353 (2020-08-25)
Checkpoint blockade immunotherapy (CBI) is effective in promoting a systemic immune response against some metastatic tumors. The reliance on the pre-existing immune environment of the tumor, however, limits the efficacy of CBI on a broad spectrum of cancers. Herein, we
Kaiyuan Ni et al.
Science advances, 6(40) (2020-10-04)
Cancer vaccines have been actively pursued to bolster antitumor immunity. Here, we designed nanoscale metal-organic frameworks (nMOFs) as locally activable immunotherapeutics to release danger-associated molecular patterns (DAMPs) and tumor antigens and deliver pathogen-associated molecular patterns (PAMPs) for in situ personalized
Alessandra Petrelli et al.
The Journal of clinical investigation, 128(10), 4669-4681 (2018-09-11)
Chronic inflammatory diseases are characterized by recurrent inflammatory attacks in the tissues mediated by autoreactive T cells. Identity and functional programming of CD8+ T cells at the target site of inflammation still remain elusive. One key question is whether, in

Protocols

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Use this protocol to for the entire immunohistochemistry (IHC) procedure through staining and visualization of specific antigens in paraffin-embedded tissue sections.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service