860336
3,3′,5,5′-Tetramethylbenzidine
≥99%
Synonym(s):
BM blue, Sure Blue TMB, TMB, TMB Blotting Plus, TMB substrate
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
Linear Formula:
[-C6H2(CH3)2-4-NH2]2
CAS Number:
Molecular Weight:
240.34
Beilstein:
2808541
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥99%
form
powder
mp
168-171 °C (lit.)
storage temp.
2-8°C
SMILES string
Cc1cc(cc(C)c1N)-c2cc(C)c(N)c(C)c2
InChI
1S/C16H20N2/c1-9-5-13(6-10(2)15(9)17)14-7-11(3)16(18)12(4)8-14/h5-8H,17-18H2,1-4H3
InChI key
UAIUNKRWKOVEES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
3,3′,5,5′-Tetramethylbenzidine is a peroxidase sensitive dye. It is a better alternative to syringaldazine for the determination of chlorine in water due to its product stability and no interference from calcium and magnesium ions.
Application
3,3′,5,5′-Tetramethylbenzidine (TMB) is a noncarcinogenic substitute (Ames test negative) for benzidine suitable as peroxidase substrate for use in ELISA procedures. The substrate produces a soluble end product that is pale blue in color and can be read spectrophotometrically at 370 or 620-650 nm. The TMB reaction may be stopped with 2 M H2SO4 (resulting in a yellow color), and read at 450 nm. A sensitive and specific reagent for the detection of blood, assay of hemoglobin, assay of peroxidases.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4 - Carc. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Colorimetric method for determination of chlorine with 3, 3?, 5, 5?-tetramethylbenzidine.
Serrat F B
Talanta, 41(12), 2091-2094 (1994)
Iron oxide nanoparticles: Hidden talent
Perez J M
Nature Nanotechnology, 2(9), 535-535 (2007)
Y Jung et al.
Mucosal immunology, 8(4), 930-942 (2015-01-08)
Eosinophils are multifunctional leukocytes that reside in the gastrointestinal (GI) lamina propria, where their basal function remains largely unexplored. In this study, by examining mice with a selective deficiency of systemic eosinophils (by lineage ablation) or GI eosinophils (eotaxin-1/2 double
Jan Pencik et al.
Nature communications, 6, 7736-7736 (2015-07-23)
Prostate cancer (PCa) is the most prevalent cancer in men. Hyperactive STAT3 is thought to be oncogenic in PCa. However, targeting of the IL-6/STAT3 axis in PCa patients has failed to provide therapeutic benefit. Here we show that genetic inactivation
Jiabin Yan et al.
Viruses, 7(3), 1429-1453 (2015-03-26)
Signaling through the Fas/Apo-1/CD95 death receptor is known to affect virus-specific cell-mediated immune (CMI) responses. We tested whether modulating the Fas-apoptotic pathway can enhance immune responses to DNA vaccination or lymphocytic choriomeningitis virus (LCMV) infection. Mice were electroporated with plasmids
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service