Skip to Content
Merck
All Photos(2)

Key Documents

39255

Sigma-Aldrich

4-Dimethylamino-4′-nitrostilbene

≥99.8% (T)

Synonym(s):

N,N-Dimethyl-4′-nitro-4-stilbenamine, DANS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CH=CHC6H4NO2
CAS Number:
Molecular Weight:
268.31
Beilstein:
1379389
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

Assay

≥99.8% (T)

form

solid

mp

256-259 °C (lit.)
256-259 °C

fluorescence

λex 500 nm; λem ~700 nm in methylene chloride

SMILES string

CN(C)c1ccc(\C=C\c2ccc(cc2)[N+]([O-])=O)cc1

InChI

1S/C16H16N2O2/c1-17(2)15-9-5-13(6-10-15)3-4-14-7-11-16(12-8-14)18(19)20/h3-12H,1-2H3/b4-3+

InChI key

NVLSIZITFJRWPY-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Dimethylamino-4′-nitrostilbene (DANS, DMANS) may be used as a proton/acid indicator and physical chemistry probe to study state energy and photoexcitation dynamics of molecules.

Other Notes

Standard for the correction of emission of fluorescence spectrometers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

I D Petsalakis et al.
The journal of physical chemistry. A, 114(17), 5580-5587 (2010-04-14)
A theoretical investigation on the electronic structure of 4-dimethylamino-4'-nitrostilbene (DANS), 4-(dicyanomethylene)-2-methyl-6-p-(dimethylamino) styryl-4H-pyran (DCM), and their protonated forms is presented in an effort to rationalize recent experimental results on the tuning of the emitted color of organic light-emitting diodes through photochemically
Antonios M Douvas et al.
Inorganic chemistry, 48(11), 4896-4907 (2009-05-02)
The capability of ammonium 18-molybdodiphosphate, (NH(4))(6)P(2)Mo(18)O(62) (Mo(18)(6-)), and ammonium 18-tungstodiphosphate, (NH(4))(6)P(2)W(18)O(62) (W(18)(6-)), to photochemically generate acid within films of a polymer with hydroxylic functional groups (namely, within poly(2-hydroxyethyl methacrylate) (PHEMA) films) is demonstrated. Upon UV irradiation, both 2:18 polyoxometalates (POMs)
Lenhart et al.
Journal of colloid and interface science, 221(1), 75-86 (2000-01-07)
Silane coupling agents are commonly applied to glass fibers to promote fiber/resin adhesion and enhance durability in composite parts. In this study, a coupling agent multilayer on glass was doped with trace levels of the dimethylaminonitrostilbene (DMANS) fluorophore. The fluorophore
Antonio Barbon et al.
Physical chemistry chemical physics : PCCP, 8(43), 5069-5078 (2006-11-09)
Two dyes (4-nitrostilbene, NST and 4-N,N-dimethylamino-4'-nitrostilbene, DANS) included in zeolites with nanometric channels and different Si : Al ratios have been photoexcited and their triplet state studied by time resolved EPR (TR-EPR). This is the first time that a TR-EPR
Takakazu Nakabayashi et al.
Journal of the American Chemical Society, 127(19), 7041-7052 (2005-05-12)
External electric field effects on state energy and photoexcitation dynamics have been examined for para-substituted and unsubstituted all-trans-diphenylpolyenes doped in a film, based on the steady-state and picosecond time-resolved measurements of the field effects on absorption and fluorescence. The substitution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service