Skip to Content
Merck
All Photos(1)

Key Documents

394475

Sigma-Aldrich

Perylene

sublimed grade, ≥99.5%

Synonym(s):

Dibenz[de,kl]anthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1911335
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

sublimed grade

Quality Level

Assay

≥99.5%

form

solid

loss

0.5 wt. % loss on heating, 232°C (TGA)

mp

276-279 °C (lit.)

λmax

436 nm

fluorescence

λem 447 nm in THF

Orbital energy

HOMO 5.3 eV 
LUMO 2.3 eV 

OLED Device Performance

ITO/NPD/ADN:Perylene/Alq3/Mg:Ag

  • Color: blue
  • Max. Luminance: 8000 Cd/m2
  • Turn-On Voltage: 3.4 V

SMILES string

c1cc2cccc3c4cccc5cccc(c(c1)c23)c45

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H

InChI key

CSHWQDPOILHKBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perylene molecules are fluorescent in nature. Ultraviolet photoemission spectroscopy and low energy electron diffraction of the structure of 4Å thick perylene dye layers adsorbed on the surface of Ru (0001) has been investigated. [1]

Application

Perylene was used in the fabrication of gold nanoparticle (AuNP) : poly(ethylene oxide) (PEO) nanocomposite thin films. [2] Perylene nanoparticles doped into poly(acrylonitrile) (PAN) were used to monitor the temperature change caused due to the nanoparticle plasmon-mediated heating within the polymer fibres and films. [3] Ordered epitaxial perylene films may be used to fabricate a multilayered electroluminescent device. [4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electroluminescence of epitaxial perylene films
Tida Y and Yanagi H
Applied Physics Letters null
Thermal Annealing of Polymer Nanocomposites via Photothermal Heating: Effects on Crystallinity and Spherulite Morphology.
Viswanath V, et al.
Macromolecules null
The growth of perylene on Ru (0001)
Honying M and Han H
J. Chem. Phys. null
Metal Nanoparticles Acting as Light?Activated Heating Elements within Composite Materials.
Maity S, et al.
Advances in Functional Materials null
Andrew Danos et al.
The journal of physical chemistry letters, 6(15), 3061-3066 (2015-08-13)
Photochemical upconversion via triplet-triplet annihilation is a promising technology for improving the efficiency of photovoltaic devices. Previous studies have shown that the efficiency of upconversion depends largely on two rate constants intrinsic to the emitting species. Here, we report that

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service