Skip to Content
Merck
All Photos(1)

Documents

D3418

Sigma-Aldrich

4-(4-Diethylaminostyryl)-1-methylpyridinium iodide

≥97%

Synonym(s):

4-Di-2-ASP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23IN2
CAS Number:
Molecular Weight:
394.29
Beilstein:
6101936
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

mp

214-216 °C (lit.)

fluorescence

λex 485 nm; λem 607 nm in methanol

SMILES string

[I-].CCN(CC)c1ccc(\C=C\c2cc[n+](C)cc2)cc1

InChI

1S/C18H23N2.HI/c1-4-20(5-2)18-10-8-16(9-11-18)6-7-17-12-14-19(3)15-13-17;/h6-15H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

WIPKWLIHFGTFQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Two-photon absorption of organic / polyoxometalate hybrid dyes
  • Acting as an acceptor for the 1D light-harvesting antenna
  • Functional live cell fluorescence imaging for pulmonary neuroepithelial body microenvironments
  • Studies of solvatochromism and dipole moments of monochromophoric styrylpyridinium dyes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 14(2), 796-808 (1994-02-01)
Remodeling of the synaptic extracellular matrix (ECM) and its dynamic relationship with nerve terminal plasticity have been demonstrated in normal frog neuromuscular junctions (NMJs) in vivo (Chen et al., 1991). Our previous work has led to a hypothesis that extension
M Hanani et al.
Journal of pediatric surgery, 28(7), 939-941 (1993-07-01)
A simple and reliable method is described for staining the nerve plexuses of the human gastrointestinal tract in whole-mounts. The staining does not require fixation and consists of a single step of 6 to 10 minutes incubation in a solution
Falk Schrödl et al.
Investigative ophthalmology & visual science, 44(9), 3705-3712 (2003-08-27)
The chemical coding of intrinsic choroidal neurons (ICNs) has features in common with extrinsic fibers (e.g., from the pterygopalatine ganglion) making it impossible to assess whether a neuronal nitric oxide synthase (nNOS)/vasoactive intestinal polypeptide (VIP)-immunoreactive nerve fiber is of intrinsic
Shih-Wei Chou et al.
Nature communications, 8(1), 2234-2234 (2017-12-23)
Detection of water motion by the lateral line relies on mechanotransduction complexes at stereocilia tips. This sensory system is comprised of neuromasts, patches of hair cells with stereociliary bundles arranged with morphological mirror symmetry that are mechanically responsive to two
Zongwei Chen et al.
Human molecular genetics, 29(12), 2004-2021 (2020-03-14)
Perception of sound is initiated by mechanically gated ion channels at the tips of stereocilia. Mature mammalian auditory hair cells require transmembrane channel-like 1 (TMC1) for mechanotransduction, and mutations of the cognate genetic sequences result in dominant or recessive heritable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service