Skip to Content
Merck
All Photos(6)

Key Documents

M1296

Sigma-Aldrich

Malonic acid

ReagentPlus®, 99%

Synonym(s):

Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein:
1751370
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor pressure

0.002 hPa ( 25 °C)

product line

ReagentPlus®

Assay

99%

form

chunks
granular
powder or crystals

bp

215 °C/18.66 hPa

mp

132-135 °C (dec.) (lit.)

solubility

1 M NaOH: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

density

1,6 g/cm3

functional group

carboxylic acid

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

General description

Malonic acid (MA), also known as propanedioic acid, is a dicarboxylic acid. The crystals of MA are triclinic at room temperature.[1][2] The oxidation of malonic acid by cerium (IV) in sulfuric acid solution has been studied.[3] The reaction kinetics of the photocatalytic decomposition of MA in aqueous suspensions of titanium dioxide (TiO2) have been described.[4]

Application

Malonic acid may be used as a cross-linking agent between corn starch and potato starch to improve its mechanical properties.[5]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Refinement of the crystal structure of malonic acid, C3H4O4.
Jagannathan NR, et al.
Journal of Chemical Crystallography, 24(1), 75-78 (1994)
On the oxidation of malonic acid by ceric ions.
Barkin S, et al.
International Journal of Chemical Kinetics, 10(6), 619-636 (1978)
The crystal structure of malonic acid.
Goedkoop JA and MacGillavry CH.
Acta Crystallographica, 10(2), 125-127 (1957)
Photocatalytic degradation of malonic acid in aqueous suspensions of titanium dioxide: an initial kinetic investigation of CO2 photogeneration.
Inel Y and Okte AN.
Journal of Photochemistry and Photobiology A: Chemistry, 96(1), 175-180 (1996)
Bogdan Belgorodsky et al.
Bioconjugate chemistry, 16(5), 1058-1062 (2005-09-22)
The preparation and characterization of the stable human serum albumin (HSA)-C3 isomer of tris-malonic acid [C60]fullerene complex is reported. Other than the anti-fullerene antibody, a stable protein-fullerene complex with a native protein has never been observed. This study may provide

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service