Skip to Content
Merck
All Photos(1)

Key Documents

24135

Sigma-Aldrich

1-Propanol

≥99% (GC), purum

Synonym(s):

Propyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2OH
CAS Number:
Molecular Weight:
60.10
Beilstein:
1098242
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99% (GC)
application(s):
microbiology
bp:
97 °C (lit.)
vapor pressure:
10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

vapor density

2.1 (vs air)

Quality Level

vapor pressure

10 mmHg ( 147 °C)
14.9 mmHg ( 20 °C)

grade

purum

Assay

≥99% (GC)

form

liquid

autoignition temp.

700 °F

expl. lim.

13.7 %

impurities

≤0.002% free acid (as C2H5COOH)
≤0.005% non-volatile matter
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.384 (lit.)

pH

8.5 (20 °C, 200 g/L)

bp

97 °C (lit.)

mp

−127 °C (lit.)

density

0.804 g/mL at 25 °C (lit.)

application(s)

microbiology

SMILES string

CCCO

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which resembles the α-relaxation and Johari-Goldstein type β-relaxation features of the supercooled liquids (nonhydrogen-bonding type). It has been tested as a substitute of the fuel for various fuel cells. Its biosynthesis from glucose using Escherichia coli strain has been proposed.

Application

1-Propanol has been employed as solvent for the synthesis of mesoporous alumina.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamics of glass-forming liquids. III. Comparing the dielectric α-and β-relaxation of 1-propanol and o-terphenyl.
Hansen C, et al.
J. Chem. Phys. , 107, 1086-1093 (1997)
Mesoporous alumina (I): comparison of synthesis schemes using anionic, cationic, and non-ionic surfactants.
Ray JC, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 100(1), 183-190 (2007)
C R Shen et al.
Metabolic engineering, 10(6), 312-320 (2008-09-09)
Production of higher alcohols via the keto-acid intermediates found in microorganism's native amino-acid pathways has recently shown promising results. In this work, an Escherichia coli strain that produces 1-butanol and 1-propanol from glucose was constructed. The strain first converts glucose
Dieuwke Sevenster et al.
Science (New York, N.Y.), 339(6121), 830-833 (2013-02-16)
Although reconsolidation opens up new avenues to erase excessive fear memory, subtle boundary conditions put constraints on retrieval-induced plasticity. Reconsolidation may only take place when memory reactivation involves an experience that engages new learning (prediction error). Thus far, it has
Anja K K Rahn et al.
Journal of agricultural and food chemistry, 61(37), 8743-8751 (2013-08-24)
Chloropropanol (CP) esters are part of an emerging group of process-induced toxicants that are considered as potential health hazards particularly in palm oil. Mass spectrometry-based methodologies for identification of CP esters in food are critical in overcoming the challenges associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service