Skip to Content
Merck
All Photos(1)

Key Documents

1.01024

Supelco

1-Propanol

for liquid chromatography LiChrosolv®

Synonym(s):

1-Propanol, n-Propyl alcohol, Propan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-746-9
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
96.5-98 °C/1013 hPa
vapor pressure:
19 hPa ( 20 °C)

vapor pressure

19 hPa ( 20 °C)

Quality Level

description

isocratic

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

360 °C

expl. lim.

2.1-19.2 % (v/v)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤2.0 mg/L

transmittance

230 nm, ≥70%
240 nm, ≥80%
270 nm, ≥98%

pH

7 (20 °C, 200 g/L in H2O)

bp

96.5-98 °C/1013 hPa

mp

-127 °C

transition temp

flash point 15 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OCCC

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Corrosion behavior of AZ91D magnesium alloy in distilled water.: This study examines the corrosion behavior of AZ91D magnesium alloy when exposed to distilled water. The research provides valuable insights into the material′s durability and the protective measures required for its use in various applications, highlighting the role of distilled water in corrosion studies (Huang and Yang, 2020).

  • Expression of concern: Metal-free green synthesis of aryl amines in magnetized distilled water: experimental aspects and molecular dynamics simulation.: Despite the expression of concern, this paper initially presented a novel approach to synthesizing aryl amines using magnetized distilled water, emphasizing the innovative applications of distilled water in green chemistry (Simpson, 2020).

  • Retraction: Metal-free green synthesis of aryl amines in magnetized distilled water: experimental aspects and molecular dynamics simulation.: This retracted article discussed the use of distilled water in the green synthesis of aryl amines, highlighting the potential and challenges of distilled water applications in chemical synthesis and analytical processes (Rowan, 2022).


Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.02 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 230 nm): ≥ 70 %
Transmission (at 240 nm): ≥ 80 %
Transmission (from 270 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An HPTLC method for rapid urea detection in milk using a TLC plate scanner and Silica Gel 60 F254 HPTLC plate.

An HPTLC method for rapid urea detection in milk using a TLC plate scanner and Silica Gel 60 F254 HPTLC plate.

An HPTLC method for rapid urea detection in milk using a TLC plate scanner and Silica Gel 60 F254 HPTLC plate.

An HPTLC method for rapid urea detection in milk using a TLC plate scanner and Silica Gel 60 F254 HPTLC plate.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service