Skip to Content
Merck
All Photos(2)

Key Documents

179787

Sigma-Aldrich

tert-Butyl methyl ether

reagent grade, ≥98%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
grade:
reagent grade
Assay:
≥98%
bp:
55-56 °C (lit.)
vapor pressure:
14.78 psi ( 55 °C)
4.05 psi ( 20 °C)
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

vapor density

3.1 (vs air)

vapor pressure

14.78 psi ( 55 °C)
4.05 psi ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

impurities

≤0.0005% peroxides (as H2O2)
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C

density

0.74 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl ether is a commonly used organic solvent.[1] It can be synthesized by acid catalyzed reaction between methanol and isobutene. It is an effective alternative to lead containing additives for enhancing the octane rating of gasoline.[2]

Application

tert-Butyl methyl ether may be used to synthesize fatty acid methyl esters (FAMEs) and glycerol tert-butyl ether via transesterification with canola oil under supercritical conditions.[3] Its addition during the polymerization of propene by stopped-flow method increases the number of active sites.[4]

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of methyl tert-butyl ether (MTBE) with Nafion.
Lien HL and Zhang WX
Journal of Hazardous Materials, 144(1), 194-199 (2007)
Relative solubility, stability, and absorptivity of lutein and β-carotene in organic solvents.
Craft NE and Soares JH.
Journal of Agricultural and Food Chemistry, 40(3), 431-434 (1992)
Eagleson M.
Concise Encyclopedia Chemistry, 154-154 (1994)
New approach of catalyst-free biodiesel production from canola oil in supercritical tert-butyl methyl ether (MTBE).
Farobie O, et al.
Fuel: The Science and Technology of Fuel and Energy, 135, 172-181 (2014)
Role of tert-Butyl Methyl Ether (TBME) as an External Donor in Propene Polymerization with Dibutyl Phthalate (DBP)-Containing MgCl2-Supported Ti Catalysts Activated with Al(i-C4H9)3.
Ohnishi R and Konakazawa T.
Macromolecular Chemistry and Physics, 205(14), 1938-1947 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service