Skip to Content
Merck
All Photos(1)

Key Documents

675407

Sigma-Aldrich

tert-Butyl methyl ether

HPLC Plus, for HPLC, GC, and residue analysis, 99.9%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC Plus
for residue analysis
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
55-56 °C (lit.)
vapor pressure:
4.05 psi

grade

HPLC Plus
for residue analysis

Quality Level

vapor density

3.1 (vs air)

vapor pressure

4.05 psi

Assay

99.9%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water

evapn. residue

≤0.0001%

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C

density

0.74 g/mL at 25 °C (lit.)

UV absorption

λ: 210 nm Amax: ≤1.00
λ: 225 nm Amax: ≤0.50
λ: 250 nm Amax: ≤0.10
λ: 300 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

suitability

suitable for residue analysis

application(s)

food and beverages

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl ether (MTBE) is a commonly used organic solvent. It can be synthesized by acid-catalyzed reaction between methanol and isobutene. It is an effective alternate to lead-containing additives for enhancing the octane rating of gasoline. The decomposition mechanism of MTBE by heat-assisted persulfate oxidation under different conditions has been reported. A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by the stopped-flow method.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid extraction by methyl-tert-butyl ether for high-throughput lipidomics.
Matyash V, et al.
Journal of Lipid Research, 49(5), 1137-1146 (2008)
Relative solubility, stability, and absorptivity of lutein and β-carotene in organic solvents.
Craft NE and Soares JH.
Journal of Agricultural and Food Chemistry, 40(3), 431-434 (1992)
Role of tert-Butyl Methyl Ether (TBME) as an External Donor in Propene Polymerization with Dibutyl Phthalate (DBP)-Containing MgCl2-Supported Ti Catalysts Activated with Al(i-C4H9)3.
Ohnishi R and Konakazawa T.
Macromolecular Chemistry and Physics, 205(14), 1938-1947 (2004)
New approach of catalyst-free biodiesel production from canola oil in supercritical tert-butyl methyl ether (MTBE).
Farobie O, et al.
Fuel: The Science and Technology of Fuel and Energy, 135, 172-181 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 154-154 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service