Skip to Content
Merck
All Photos(1)

Key Documents

1.01845

Supelco

tert-Butyl methyl ether

for liquid chromatography LiChrosolv®

Synonym(s):

tert-Butyl methyl ether, MTBE, MTB, 2-Methoxy-2-methylpropane, Methyl tert-butyl ether, TBME

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
MDL number:
UNSPSC Code:
12191502
EC Index Number:
216-653-1
NACRES:
NA.03
technique(s):
HPLC: suitable
application(s):
PFAS testing
bp:
55.3 °C/1013 hPa
vapor pressure:
268 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

268 hPa ( 20 °C)

Quality Level

description

isocratic

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

460 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-8.5 % (v/v)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤2.0 mg/L

transmittance

240 nm, ≥60%
255 nm, ≥85%
280 nm, ≥98%

kinematic viscosity

0.409 cSt(40 °C)

bp

55.3 °C/1013 hPa

mp

-108.6 °C

transition temp

flash point -28 °C

solubility

42 g/L

density

0.74 g/cm3 at 20 °C

application(s)

PFAS testing

storage temp.

2-30°C

SMILES string

O(C(C)(C)C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

General description

LiChrosolv® solvents provide high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.

Application

Intended for HPLC and UHPLC-MS applications.

Used as an extraction solvent to analyze nucleotides of Drosophila gut microbiome in flies prior to LC-MS analysis.[1]

Features and Benefits

  • suitability tested and specified for UHPLC-MS and UHPLC-UV: for analytical flexibility
  • specified quality in positive and negative in positive and negative electrospray ionization (ESI) and atmospheric-pressure chemical ionization-mass spectrometry (APCI-MS) for lowest detection limits and confidence in analyses for all important MS modes (test 1)
—ESI/APCI (+) < 2 ppb
—ESI/APCI (-) < 10 ppb
  • lowest impurity profile: for interference-free baselines (test 2)
  • Microfiltered through 0.2 μm filter (test 3) to provide
—a long lifespan to filters and mechanical parts of HPLC systems
—reduced risk of column clogging
  • packed in borosilicate glass bottles for minimized metal ion contaminations
  • lowest levels of trace metal impurities for a minimum metal ion adduct formation
—<5 ppb
  • lowest level of polyethylene glycol (PEG) impurities in UHPLC-MS solvent lineup (PEG S/N signal-to-noise-ratio < 50)

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.02 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 240 nm): ≥ 60 %
Transmission (at 255 nm): ≥ 85 %
Transmission (from 280 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocol for studying microbiome impact on host energy and reproduction in Drosophila
Gnainsky Y, et al.
STAR protocols, 3(2), 101253-101253 (2022)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service