Skip to Content
Merck
All Photos(2)

Key Documents

T0886

Sigma-Aldrich

Triethylamine

≥99%

Synonym(s):

TEA, N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21
bp:
88.8 °C (lit.)
vapor pressure:
51.75 mmHg ( 20 °C)

vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

593 °F

shelf life

3 yr

expl. lim.

8 %

impurities

≤0.5% water (Karl Fischer)

refractive index

n20/D 1.401 (lit.)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triethylamine has been used:
  • as a hydrogen donor for the polymerization of various monomers
  • to catalyze silanization
  • in the synthesis of the Cy3-Alexa647 heterodimer
  • in the synthesis of methacrylated solubilized decellularized cartilage

Biochem/physiol Actions

Triethylamine is known to drive polymerization reaction. It acts as a source of carbon and nitrogen for bacterial cultures. Triethylamine is used in pesticides. Triethylamine can serve as an organic solvent.

also commonly purchased with this product

suggested gloves for splash protection

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of tissue processing on bioactivity of cartilage matrix-based hydrogels encapsulating osteoconductive particles.
Townsend JM, et al.
Biomedical Materials (Bristol, England), 13(3) (2018)
Wide-field in vivo background free imaging by selective magnetic modulation of nanodiamond fluorescence.
Sarkar SK, et al.
Biomedical Optics Express, 5(4), 1190-1202 (2014)
Synthesis and characterization of poly(vinylchloride) type macrophotoinitiator comprising side-chain thioxanthone via click chemistry
Akat H and Ozkan M
Express Polymer Letters, 5(4), 318?326-318?326 (2011)
The biodegradation pathway of triethylamine and its biodegradation by immobilized Arthrobacter protophormiae cells
Cai T, et al.
Journal of Hazardous Materials, 186(1), 59-66 (2011)
RESOLFT nanoscopy with photoswitchable organic fluorophores.
Kwon J, et al.
Scientific reports (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service