Skip to Content
Merck
All Photos(2)

Key Documents

E3893

Sigma-Aldrich

(−)-Epicatechin gallate

≥98% (HPLC), from green tea

Synonym(s):

(−)-cis-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,7-Pentahydroxyflavane 3-gallate, 3-O-Galloylepicatechin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O10
CAS Number:
Molecular Weight:
442.37
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

green tea

Assay

≥98% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4ccc(O)c(O)c4

InChI

1S/C22H18O10/c23-11-6-14(25)12-8-19(32-22(30)10-4-16(27)20(29)17(28)5-10)21(31-18(12)7-11)9-1-2-13(24)15(26)3-9/h1-7,19,21,23-29H,8H2/t19-,21-/m1/s1

InChI key

LSHVYAFMTMFKBA-TZIWHRDSSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Epicatechin gallate is one of the major polyphenols present in green tea. It belongs to the catechin gallates group of organic compounds. ECG has a chemical structure like that of (−)-epigallocatechin gallate (EGCG), the other major polyphenol found in green tea.

Application

(−)-Epicatechin gallate has been used:
  • in cell proliferation assays on the human brain microvascular endothelial cells (HBMVECs) to understand its effectiveness against anti-ischemic/reperfusion injury
  • as an inhibitor in Human Salivary α-Amylase (HAS) and α-Glucosidase (AGH) in inhibition assays
  • may be used as a cancer chemopreventive polyphenol in two human colorectal cell lines HCT-116 and SW-480
  • as an inhibitor to in vitro collagenase and elastase enzymes
  • as a supplement in spermatozoa incubation to check its effectiveness against oxidative stress

Biochem/physiol Actions

(−)-Epicatechin gallate (ECG), a potent free-radical scavenger has an inhibitory effect on collagenase and elastase enzymes making it an anti-aging agent. It displays antiproliferative effects on human colorectal cells and hence is a chemo-preventative agent. It influences cell survival, preventing apoptosis or autophagy by promoting cell proliferation in oxygen-glucose deprived human brain microvascular endothelial cells. Being structurally and functionally similar to EGCG, ECG might be involved in balancing the mammalian target of rapamycin (mTOR)-AMP-activated protein kinase (AMPK) pathways in endoplasmic reticulum stress. It prevents cell death in this scenario by showing resistance against oxygen-deprived cells. ECG may contribute in preventing oxidative stress. It effectively inhibits secretory sphingomyelinase in many disease states and provides protection for human spermatozoa in Assisted reproductive technology (ART).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee Wah Koh et al.
Journal of agricultural and food chemistry, 58(1), 148-154 (2010-01-07)
Current work investigated the ability of different tea (green, oolong and black teas) in inhibiting human salivary alpha-amylase (HSA) and mammalian alpha-glucosidase (AGH). The inhibitory profiles were correlated to their major polyphenol content (theaflavins and catechins). The fully fermented black
Bing Fu et al.
Oxidative medicine and cellular longevity, 2019, 7824684-7824684 (2019-04-10)
Green tea is one of the most beverages with antioxidants and nutrients. As one of the major components of green tea, (-)-epicatechin gallate (ECG) was evaluated for its antioxidative properties in the present study. Cell proliferation assay, tube formation, cell
Marianna Holczer et al.
Oxidative medicine and cellular longevity, 2018, 6721530-6721530 (2018-04-11)
The maintenance of cellular homeostasis is largely dependent on the ability of cells to give an adequate response to various internal and external stimuli. We have recently proposed that the life-and-death decision in endoplasmic reticulum (ER) stress response is defined
Keiko Kobayashi et al.
Journal of nutritional science and vitaminology, 62(2), 123-129 (2016-06-07)
Sphingomyelinases (SMases) are key enzymes involved in many diseases which are caused by oxidative stress, such as atherosclerosis, diabetes mellitus, nonalcoholic fatty liver disease, and Alzheimer's disease. SMases hydrolyze sphingomyelin to generate ceramide, a well-known pro-apoptotic lipid. SMases are classified
Wanda C Reygaert
BioMed research international, 2018, 9105261-9105261 (2018-08-15)
Green tea is one of the most popular drinks consumed worldwide. Produced mainly in Asian countries from the leaves of the Camellia sinensis plant, the potential health benefits have been widely studied. Recently, researchers have studied the ability of green

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service