Skip to Content
Merck
All Photos(1)

Documents

08108

Supelco

(−)-Epigallocatechin

analytical standard

Synonym(s):

(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3cc(O)c(O)c(O)c3

InChI

1S/C15H14O7/c16-7-3-9(17)8-5-12(20)15(22-13(8)4-7)6-1-10(18)14(21)11(19)2-6/h1-4,12,15-21H,5H2/t12-,15-/m1/s1

InChI key

XMOCLSLCDHWDHP-IUODEOHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Epigallocatechin (EGC) belongs to the tea catechins group of polyphenols.

Application

EGC may be used as an analytical standard for the determination of EGC in:
  • Fruit juices by high-performance liquid chromatography with diode array detection (HPLC-DAD) combined with electrospray ionization (ESI) and triple quadrupole mass spectrometry.
  • Green tea standard reference materials (SRMs) by LC coupled with particle beam/electron ionization mass spectrometry (LC-PB/EIMS).
  • Tea extracts by HPLC with electrochemical detection (ECD) and in plant-based foods by HPLC-DAD.

Biochem/physiol Actions

Green tea polyphenol. Anticancer agent. Lower antioxidant activity than EGCG.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general analytical strategy for the characterization of phenolic compounds in fruit juices by high-performance liquid chromatography with diode array detection coupled to electrospray ionization and triple quadrupole mass spectrometry
Abad-Garcia, B, et al.
Journal of Chromatography A, 1216(28), 5398-5415 (2009)
Determination of catechins and caffeine in proposed green tea standard reference materials by liquid chromatography-particle beam/electron ionization mass spectrometry (LC-PB/EIMS)
Castro J, et al.
Talanta, 82(5), 1687-1695 (2010)
Pharmacokinetics of tea catechins after ingestion of green tea and (-)-epigallocatechin-3-gallate by humans: formation of different metabolites and individual variability
Lee MJ, et al.
Cancer Epidemiology, Biomarkers & Prevention, 11(10), 1025-1032 (2002)
Liquid chromatographic method for the separation and quantification of prominent flavonoid aglycones
Merken HM and Beecher GR
Journal of Chromatography A, 897(1-2), 177-184 (2000)
Simultaneous determination of twelve tea catechins by high-performance liquid chromatography with electrochemical detection
Sano M, et al.
Analyst, 126(6), 816-820 (2001)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service