Skip to Content
Merck
All Photos(3)

Documents

E4018

Sigma-Aldrich

(−)-Epicatechin

≥98% (HPLC), from green tea

Synonym(s):

(−)-cis-3,3′,4′,5,7-Pentahydroxyflavane, (2R,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
Beilstein:
92760
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.77

biological source

green tea

Assay

≥98% (HPLC)

mp

240 °C (dec.) (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15-/m1/s1

InChI key

PFTAWBLQPZVEMU-UKRRQHHQSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Epicatechin is a flavanol, that belongs to the class of secondary, polyphenolic plant metabolites.

Application

(-)-Epicatechin has been used:
  • to examine its biological effects in cell lines from human gender-specific cancers
  • to study its antiviral activity
  • to test its ability to regenerate this enzymatic activity both in vitro at the isolated MPO−H2O2−Cl- system and ex vivo in human polymorphonuclear leukocytes (PMNs)

Biochem/physiol Actions

This antioxidant is found in chocolate. (-)-Epicatechin is linked with cardiovascular effects. It is considered as a dependable standard due to its availability in cacao seeds and cocoa products. The halogenating activity of myeloperoxidase can be restored by (-)-epicatechin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Divergent antiviral effects of bioflavonoids on the hepatitis C virus life cycle
Khachatoorian R, et al.
Virology, 346-355 (2012)
Epicatechins purified from green tea (Camellia sinensis) differentially suppress growth of gender-dependent human cancer cell lines
Ravindranath M H, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 3(2), 237-247 (2006)
Fluorescent detection of (-)-epicatechin in microsamples from cacao seeds and cocoa products: Comparison with Folin-Ciocalteu method
Ramirez-Sanchez I, et al.
Journal of Food Lipids, 23(8), 790-793 (2010)
(-)-Epicatechin mediates beneficial effects of flavanol-rich cocoa on vascular function in humans
Schroeter H, et al.
Proceedings of the National Academy of Sciences of the USA, 103(4), 1024-1029 (2006)
(-)-Epicatechin regenerates the chlorinating activity of myeloperoxidase in vitro and in neutrophil granulocytes
Flemmig J, et al.
Journal of Inorganic Biochemistry, 84-91 (2014)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service