Skip to Content
Merck
All Photos(1)

Key Documents

74156

Supelco

Nonacosane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)27CH3
CAS Number:
Molecular Weight:
408.79
Beilstein:
1724922
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

286 °C/15 mmHg (lit.)

mp

63-66 °C (lit.)
63-66 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C29H60/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-29H2,1-2H3

InChI key

IGGUPRCHHJZPBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonacosane,an aliphatic hydrocarbon, is important constituent of the leaf oil of Syzygium cumini tree, possessing anti-bacterial, anti-inflammatory and anti-ulcergenic properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neutral components in the leaves and seeds of Syzygium cumini
Kumar.A, et al.
African Journal of Pharmacy and Pharmacology, 3(11), 560-561 (2009)
Eoin P Coll et al.
The journal of physical chemistry. B, 111(30), 8702-8704 (2007-07-05)
Plant wax contains long-chain alkanes and related components which are transported to the surface of the plant by specialized ABC transporter proteins. Here, we determine the distribution and conformation of three wax components, nonacosane, nonacosan-15-one, and nonacosan-15-ol, using unbiased and
[Constituents of the Lipophilic fraction of the Apocynum venetum leaf].
P F Jiang
Zhong yao tong bao (Beijing, China : 1981), 10(5), 30-31 (1985-05-01)
Supercritical fluid extraction of triterpenes and aliphatic hydrocarbons from olive tree derivatives
Issaoui A
Arabian Journal of Chemistry, 10, S3967-S3973 (2017)
Yan-Zheng Zhang et al.
Food research international (Ottawa, Ont.), 116, 362-369 (2019-02-06)
The eastern honeybee Apis cerana and the western honeybee Apis mellifera are the two most economically valuable honeybee species used in apiculture. In market, the price of Apis cerana honey (ACH) is usually several times higher than that of Apis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service