Skip to Content
Merck
All Photos(2)

Documents

51560

Sigma-Aldrich

Heptacosane

≥98.0% (GC)

Synonym(s):

n-Heptacosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)25CH3
CAS Number:
Molecular Weight:
380.73
Beilstein:
1706242
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

bp

270 °C/15 mmHg (lit.)

mp

58-60 °C (lit.)
58-60 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C27H56/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-27H2,1-2H3

InChI key

BJQWYEJQWHSSCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ordered phases in n-heptacosane (C27H56). Structure determination of the Mdci phase.
Rajabalee F, et al.
Chemistry of Materials, 14(10), 4081-4087 (2002)
STM study of molecular adsorption on single-wall carbon nanotube surface.
Ago H, et al.
Chemical Physics Letters, 383(5), 469-474 (2004)
Distribution and thermophysical properties of n-heptacosane molecules confined in carbon nanotube.
Tong X, et al.
Phase Transitions, 1-10 (2015)
Christian Zorzetto et al.
Fitoterapia, 100, 95-109 (2014-12-03)
In the present work we carried out a phytochemical and biological investigation on three Hypericum species, i.e. Hypericum reflexum, Hypericum canariense and Hypericum grandifolium, from the Canary Islands where they are traditionally used as diuretic, wound healing, vermifuge, sedative and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service