Skip to Content
Merck
All Photos(1)

Documents

N28906

Sigma-Aldrich

Nonadecane

99%

Synonym(s):

n-Nonadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17CH3
CAS Number:
Molecular Weight:
268.52
Beilstein:
1742892
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.27 (vs air)

vapor pressure

1 mmHg ( 133.2 °C)

Assay

99%

form

crystalline
liquid
waxy solid

autoignition temp.

446 °F

bp

330 °C (lit.)

mp

30-34 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCC

InChI

1S/C19H40/c1-3-5-7-9-11-13-15-17-19-18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3

InChI key

LQERIDTXQFOHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonadecane is generally used as a subject in positron annihilation lifetime spectroscopy (PALS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Positronium in the n-alkane binary mixture: Nonadecane-heneicosane.
Zgardzinska B, et al.
Chemical Physics Letters, 470(1-3), 72-74 (2009)
Positron annihilation and phase transitions in argon intercalated n-nonadecane.
Zgardzinska B, et al.
Chemical Physics, 335(1), 1-6 (2007)
Positronium in alkanes. From neat nonadecane to wax.
Zgardzinska B and Goworek T
Chemical Physics Letters, 547(1-3), 35-37 (2012)
Nickel-Catalyzed Suzuki Polycondensation for Controlled Synthesis of Ester-Functionalized Conjugated Polymers.
Qiu Y, et al.
Macromolecules, 49(13), 4757-4762 (2016)
Stefano Colazza et al.
Journal of chemical ecology, 33(7), 1405-1420 (2007-05-18)
Contact kairomones from adult southern green stink bugs, Nezara viridula (L.) (Heteroptera: Pentatomidae) that elicit foraging behavior of the egg parasitoid Trissolcus basalis (Wollaston) were investigated in laboratory experiments. Chemical residues from tarsi and scutella of N. viridula induced foraging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service