All Photos(1)
About This Item
Linear Formula:
CH3(CH2)24CH3
CAS Number:
Molecular Weight:
366.71
Beilstein:
1705609
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
mp
55-58 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCCCCCCCCCCCC
InChI
1S/C26H54/c1-3-5-7-9-11-13-15-17-19-21-23-25-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-26H2,1-2H3
InChI key
HMSWAIKSFDFLKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Hexacosane is one of the important component of the essential oil of Moringa oleifera (Moringaceae).
Application
Hexacosane can be used:
- To prepare sulfonated hexacosanes, which are used as phase-change materials in the latent heat thermal energy storage process.
- As a model n-alkane to study the solid-liquid equilibrium of binary mixtures in N,N-dimethylacetamide and N,N-dimethylformamide.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Solid-liquid equilibria of long chain n-alkanes (C18-C28) in n, n-dimethylacetamide
Sekhar GC, et al.
Fluid Phase Equilibria, 201, 219-231 (2002)
Enthalpy of melting and solidification of sulfonated paraffins as phase change materials for thermal energy storage
Alkan C
Thermochimica Acta, 451, 126-130 (2006)
Benjamin Savage et al.
Journal of chemical ecology, 47(7), 628-641 (2021-06-24)
Cuticular hydrocarbons (CHCs) are important, multi-function components of the insect epicuticle. In Drosophila spp., CHCs provide protection from desiccation and serve as semiochemicals for both intra- and interspecific communication. We developed a non-lethal method for the modification of Drosophila CHCs
Solid-Liquid Equilibria of n-Alkanes in N,N-Dimethylformamide
Venkatesu P, et al.
Journal of Chemical and Engineering Data, 45, 177-181 (2000)
Tatiana Marrufo et al.
Molecules (Basel, Switzerland), 18(9), 10989-11000 (2013-09-12)
The antioxidant capacity and antimicrobial activity of the essential oil of Moringa oleifera (Moringaceae) grown in Mozambique was investigated. The chemical composition was studied by means of GC and GC-MS analysis. Hexacosane (13.9%), pentacosane (13.3%) and heptacosane (11.4%) were the
Protocols
Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service