Skip to Content
Merck
All Photos(2)

Key Documents

73569

Supelco

2-Methyl-4-isothiazolin-3-one

analytical standard

Synonym(s):

2-Methyl-3(2H)-isothiazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NOS
CAS Number:
Molecular Weight:
115.15
Beilstein:
606203
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GCF)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤13.0% water (Karl Fischer)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CN1SC=CC1=O

InChI

1S/C4H5NOS/c1-5-4(6)2-3-7-5/h2-3H,1H3

InChI key

BEGLCMHJXHIJLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

2-Methyl-4-isothiazolin-3-one may be used as an antimicrobial preservative for the analysis of Ochratoxin A in spices using clean-up tandem immunoassay columns followed by confirmation by high performance liquid chromatography technique and in non-formalin products for wallpaper adhesives using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of a mixture of chloromethylisothiazolinone and methylisothiazolinone on peripheral airway dysfunction in children
Cho J-H, et al.
PLoS ONE, 2(4) (2017)
Approach for ochratoxin A fast screening in spices using clean-up tandem immunoassay columns with confirmation by high performance liquid chromatography?tandem mass spectrometry (HPLC?MS/MS)
Goryacheva.Y.I, et al.
Analytica Chimica Acta, 577(1), 38-45 (2006)
Simultaneous Determination of Ultraviolet Absorbers and Antibacterial Agents in Textiles by Ultra-High Performance Liquid Chromatography/Orbitrap High Resolution Mass Spectrometry
Wang C, et al.
World Journal of Engineering and Technology, 5(01), 1-1 (2017)
Determination of antimicrobial agents in non-formalin adhesives for wallpaper
Nakashima H, et al.
Journal of Health Science, 46(6), 447-454 (2000)
Johannes Geier et al.
Contact dermatitis, 67(6), 334-341 (2012-09-22)
The frequency of sensitization to methylchloroisothiazolinone (MCI)/ methylisothiazolinone (MI) observed in the Information Network of Departments of Dermatology (IVDK) was constantly around 2.1% from 1998 to 2009. After that, it increased to 3.9% in 2011, paralleled by an increase in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service