Skip to Content
Merck
All Photos(2)

Key Documents

75169

Supelco

1,2-Benzisothiazol-3(2H)-one

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NOS
CAS Number:
Molecular Weight:
151.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤15% water

mp

152-157 °C
154-158 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1nsc2ccccc12

InChI

1S/C7H5NOS/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)

InChI key

DMSMPAJRVJJAGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Benzisothiazol-3(2H)-one belongs to the chemical class of isothiazolinones.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Application

The analytical standards can also be used as follows:

  • Determination of benzisothiazolinone in five soil samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) in positive electrospray ionization mode

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and anti-bacterial activity of a library of 1, 2-benzisothiazol-3 (2H)-one (BIT) derivatives amenable of crosslinking to polysaccharides
Viani F, et al.
Tetrahedron, 73(13), 1745-1761 (2017)
William L Jorgensen et al.
Bioorganic & medicinal chemistry letters, 21(15), 4545-4549 (2011-07-02)
Substituted N-phenylbenzisothiazolones have been investigated as inhibitors of the tautomerase activity of the proinflammatory cytokine MIF (macrophage migration inhibitory factor). Numerous compounds were found to possess antagonist activity in the low micromolar range with the most potent being the 6-hydroxy
Occupational allergic contact dermatitis caused by 1,2-benzisothiazolin-3-one in a varnish maker, followed by sensitization to benzalkonium chloride in Oilatum Plus bath additive.
S L Walker et al.
Contact dermatitis, 50(2), 104-105 (2004-05-07)
Alan Rolfe et al.
Journal of combinatorial chemistry, 11(4), 732-738 (2009-06-10)
A sequential three-component synthesis of functionalized benzisothiazoline-3-acetic acid 1,1-dioxides utilizing a domino Heck-aza-Michael pathway is reported. This one-pot procedure rapidly assembles functionalized benzylsulfonamides, which undergo a palladium-catalyzed, domino, Heck-aza-Michael transformation in an experimentally straightforward manner. This attractive protocol has been
Allergic contact dermatitis to 1,2-benzisothiazolin-3-one in the water softener manufacturing industry.
H Reddy et al.
Contact dermatitis, 61(3), 184-185 (2009-09-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service