Skip to Content
Merck
All Photos(1)

Key Documents

84822

Sigma-Aldrich

Bis(2-ethylhexyl) sebacate

≥97.0% (GC)

Synonym(s):

Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
426.67
Beilstein:
1806504
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.450 (lit.)

bp

212 °C/1 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2-ethylhexyl) sebacate is commonly used as a pressure transmitting fluid (PTF) in hydraulically operated devices, pressure measuring instruments and hydraulic pressure balances.[1] It is also a plasticizing solvent mediator for preparing poly(vinyl chloride) (PVC) based membranes.[2]

Storage Class Code

10 - Combustible liquids

WGK

awg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental density and viscosity measurements of di (2ethylhexyl) sebacate at high pressure.
Paredes X, et al.
The Journal of Chemical Thermodynamics, 44(1), 38-43 (2012)
Determination of complex formation constants of 18 neutral alkali and alkaline earth metal ionophores in poly (vinyl chloride) sensing membranes plasticized with bis (2-ethylhexyl) sebacate and o-nitrophenyloctylether.
Qin Y, et al.
Analytica Chimica Acta, 421(2), 207-220 (2000)
Yihang Chen et al.
Lab on a chip, 20(22), 4205-4214 (2020-10-14)
Microchannels in hydrogels play an essential role in enabling a smart contact lens. However, microchannels have rarely been created in commercial hydrogel contact lenses due to their sensitivity to conventional microfabrication techniques. Here, we report the fabrication of microchannels in
J S Ilowite et al.
Archives of environmental health, 44(4), 267-273 (1989-07-01)
Mucociliary clearance was compared in three groups of normal subjects; each group inhaled a different type of aerosol (sebecate, iron oxide, and aqueous) labelled with 99Tc, to determine the relationship between deposition pattern and the subsequent total lung clearance. Standardization
P J Anderson et al.
The American review of respiratory disease, 144(3 Pt 1), 649-654 (1991-09-01)
How variable is the deposition of inhaled methacholine (MCH) in the respiratory tract during a challenge test? Does this variability contribute to the variability of airway responsiveness? To examine these questions we estimated the deposition of polydisperse MCH droplets by

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service