Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Methyl-4-isothiazolin-3-one
Synonym(s):
2-Methyl-3(2
H
)-isothiazolone
Empirical Formula (Hill Notation):
C
4
H
5
NOS
CAS Number:
2682-20-4
Molecular Weight:
115.15
EC Number:
220-239-6
Products
Product Category
antioxidants (1)
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
725765
2-Methyl-4-isothiazolin-3-one,
95%
Expand
View Pricing
73569
2-Methyl-4-isothiazolin-3-one,
analytical standard
Expand
View Pricing