Skip to Content
Merck
All Photos(2)

Key Documents

45306

Supelco

Ethoprophos

PESTANAL®, analytical standard

Synonym(s):

O-Ethyl S,S-dipropyl phosphorodithioate, Ethoprofos, Ethoprop

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19O2PS2
CAS Number:
Molecular Weight:
242.34
Beilstein:
8139788
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCSP(=O)(OCC)SCCC

InChI

1S/C8H19O2PS2/c1-4-7-12-11(9,10-6-3)13-8-5-2/h4-8H2,1-3H3

InChI key

VJYFKVYYMZPMAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enric Pellicer-Castell et al.
Talanta, 189, 560-567 (2018-08-09)
A novel sorbent based on a UVM-7 mesoporous silica doped with Ti has been synthesized and used for solid-phase extraction of several organophosphorus pesticides in environmental water samples followed by gas chromatography coupled to a nitrogen-phosphorus selective detector. Thus, mesoporous
Nor Izzati Fikrah Aris et al.
Royal Society open science, 7(3), 192050-192050 (2020-04-10)
Superhydrophilic graphene oxide/electrospun cellulose nanofibre (GO/CNF) was synthesized, characterized and successfully used in a solid-phase membrane tip adsorption (SPMTA) as an adsorbent towards a simultaneous analysis of polar organophosphorus pesticides (OPPs) in several food and water samples. Separation, determination and
Lobna Hajji-Hedfi et al.
Environmental science and pollution research international, 25(18), 17278-17282 (2018-04-14)
Pot and greenhouse trials were conducted for the management of root-knot nematode, Meloidogyne javanica, infestation in tomato. Growth parameters, gall index, soil, and root nematode populations were measured to assess the effect of a novel bio-pesticide (Dazitol®), made from mustard
Ross Maciejewski et al.
Public health reports (Washington, D.C. : 1974), 123(3), 333-342 (2008-11-15)
This study utilized the electronic medical records of six veterinary hospitals (operated by Banfield, The Pet Hospital) in the vicinity of Fairburn, Georgia, to assess the health of dogs and cats following the unintentional release of propyl mercaptan from a
D G Karpouzas et al.
Journal of applied microbiology, 89(1), 40-48 (2000-08-17)
Two strains of Pseudomonas putida (epI and epII), isolated previously from ethoprophos-treated soil, were able to degrade ethoprophos (10 mg 1(-1)) in a mineral salts medium plus nitrogen (MSMN) in less than 50 h with a concurrent population growth. Addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service