Skip to Content
Merck
All Photos(1)

Key Documents

31733

Supelco

Emamectin benzoate

PESTANAL®, analytical standard

Synonym(s):

(4′’R)-4″-Deoxy-4″-(methylamino)avermectin B1 benzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

[N+H2]([C@H]2[C@@H](O[C@H](C[C@@H]2OC)O[C@H]3[C@@H](O[C@H](C[C@@H]3OC)O[C@H]4[C@H](\C=C\C=C5\[C@@]6([C@H](OC\5)[C@@H](C(=C[C@H]6C(=O)O[C@@H]7C[C@]8(O[C@@H](C7)C\C=C\4/C)O[C@@H]([C@H](C=C8)C)[C@H](CC)C)C)O)O)C)C)C)C.[O-]C(=O)c1ccccc1

InChI

1S/C49H75NO13.C7H6O2/c1-12-26(2)44-29(5)18-19-48(63-44)24-35-21-34(62-48)17-16-28(4)43(27(3)14-13-15-33-25-56-46-42(51)30(6)20-36(47(52)59-35)49(33,46)53)60-40-23-38(55-11)45(32(8)58-40)61-39-22-37(54-10)41(50-9)31(7)57-39;8-7(9)6-4-2-1-3-5-6/h13-16,18-20,26-27,29,31-32,34-46,50-51,53H,12,17,21-25H2,1-11H3;1-5H,(H,8,9)/b14-13+,28-16+,33-15+;/t26-,27-,29-,31-,32-,34+,35-,36-,37-,38-,39-,40-,41-,42+,43-,44+,45-,46+,48+,49+;/m0./s1

InChI key

GCKZANITAMOIAR-XWVCPFKXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Emamectin benzoate is a macrocyclic insecticide found to be stable aqueous hydrolysis. Avermectin B1 can be the starting material synthesizing emamectin benzoate. In some occasions, it is used as acaricide for vegetables, fruits and cereals.

Application

Emamectin benzoate may be used as pesticide standard in supercritical fluid chromatography/tandem mass spectrometry (SFC/MS/MS) method for pesticide residue determination from brown rice, onion, and spinach samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 1 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an enzyme-linked immunosorbent assay for residue analysis of the insecticide emamectin benzoate in agricultural products.
Ishibashi M
Journal of Chromatography A, 1266, 143-148 (2012)
Roy Beryl Julinta et al.
Environmental toxicology and pharmacology, 75, 103348-103348 (2020-02-08)
Emamectin benzoate (EB) premix top-coated onto feed is extensively used to treat ectoparasitic crustacean infestations in aquaculture. This study evaluated the safety of EB-dosing in Nile tilapia Oreochromis niloticus at the recommended dose and dosage of 50 μg/kg biomass/day for 7
Development of an enzyme-linked immunosorbent assay for residue analysis of the insecticide emamectin benzoate in agricultural products.
Kondo M
Journal of Agricultural and Food Chemistry, 57(2), 359-364 (2009)
Terence Robert Roberts, David Herd Hutson
Metabolic Pathways of Agrochemicals: Herbicides and plant growth regulators;, 89-89 (2007)

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service