Skip to Content
Merck
All Photos(1)

Key Documents

45487

Supelco

Fenitrothion

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12NO5PS
CAS Number:
Molecular Weight:
277.23
Beilstein:
8983553
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(c(C)c1)[N+]([O-])=O

InChI

1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3

InChI key

ZNOLGFHPUIJIMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Amandine Guidez et al.
Memorias do Instituto Oswaldo Cruz, 115, e200313-e200313 (2021-02-04)
Aedes aegypti is the sole vector of urban arboviruses in French Guiana. Overtime, the species has been responsible for the transmission of viruses during yellow fever, dengue, chikungunya and Zika outbreaks. Decades of vector control have produced resistant populations to
A Larki
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 1-5 (2016-09-03)
In this work, the intrinsic colorimetric property of carbon dots (CDs) was utilized for the determination of fenitrothion by applying dispersive liquid-liquid microextraction (DLLME) method. Label free CDs are extracted into carbon tetrachloride via assistance of trioctylmethylammonium chloride (Aliquat 336)
Roberto Sanchirico et al.
Journal of hazardous materials, 199-200, 390-400 (2011-12-06)
The thermal decomposition of Fenitrothion [phosphorothioic acid O,O-diethyl O-(3-methyl-4-nitrophenyl) ester] was investigated. Results obtained by different scale calorimetric techniques show that the thermal decomposition of Fenitrothion involves two main steps. Intermediate and final thermal degradation products formed during isothermal and
Kuppusamy Kanagaraj et al.
Biosensors & bioelectronics, 35(1), 452-455 (2012-03-20)
A unique, efficient, highly sensitive and selective fluorescent chemosensor for fenitrothion has been reported for the first time using per-6-amino-β-cyclodextrin:Eu(III) complex. Among the various pesticides, the sensitivity response is found to be in the order, fenitrothion>quinalphos>methylparathion>parathion>methylparaoxon>paraoxon>fenchlorphos>profenofos>malathion. A detection limit as
Cristiana Oliveira et al.
Chemosphere, 90(3), 936-944 (2012-07-25)
The aim of this study was to develop two behavioral tests (swimming velocity and avoidance behavior) specific for the common prawn, Palaemon serratus, and to investigate the effects of sublethal concentrations of fenitrothion on behavior and on several biomarkers. In

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service