Skip to Content
Merck
All Photos(1)

Key Documents

442829

Supelco

Triphenyl phosphate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
CAS Number:
Molecular Weight:
326.28
Beilstein:
1888236
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

1.3 mmHg ( 200 °C)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

244 °C/10 mmHg (lit.)

mp

48-50 °C (lit.)

solubility

H2O: 0.0019 g/L at 20 °C

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

428.0 °F - closed cup

Flash Point(C)

220 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Amer El-Batta et al.
The Journal of organic chemistry, 72(14), 5244-5259 (2007-06-15)
Water is demonstrated to be an effective medium for the Wittig reaction over a wide range of stabilized ylides and aldehydes. Despite sometimes poor solubility of the reactants, good chemical yields normally ranging from 80 to 98% and high E-selectivities
Rainer Hoenicke et al.
Environmental research, 105(1), 132-144 (2007-03-06)
While over seven million organic and inorganic compounds that have been indexed by the American Chemical Society's Chemical Abstracts Service in their CAS Registry are commercially available, most pollution monitoring programs focus only on those chemical stressors for which regulatory
Chunsheng Liu et al.
Aquatic toxicology (Amsterdam, Netherlands), 128-129, 147-157 (2013-01-12)
Tris(1,3-dichloro-2-propyl) phosphate (TDCPP) and triphenyl phosphate (TPP) are frequently detected in biota, including fish. However, knowledge of the toxicological and molecular effects of these currently used flame retardants is limited. In the present study, an in vivo screening approach was
Felix D Guerrero et al.
Journal of medical entomology, 43(5), 896-901 (2006-10-05)
The horn fly, Haematobia irritans irritans (L.) (Diptera: Muscidae), has become a problem for Brazilian cattle producers even though its introduction into Brazil is relatively recent. Failure to control this cattle pest is becoming a concern, and horn fly populations
Yasunori Kawagoshi et al.
Chemosphere, 48(2), 219-225 (2002-07-16)
Degradation of organophosphoric esters (OPEs) in leachate from a sea-based solid waste disposal site was investigated by laboratory experiment. Aryl-phosphates, tricresyl phosphate and triphenyl phosphate, in leachate rapidly decreased to less than the detection limit within 20 days under aerobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service