Skip to Content
Merck
All Photos(1)

Key Documents

MAK501

Sigma-Aldrich

Glucose Oxidase Activity Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Glucose Oxidase Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes

shipped in

wet ice

storage temp.

−20°C

General description

Glucose oxidase catalyzes the oxidation of glucose from D-glucose to D-glucono-δ-lactone. Physiologically, it aids in the breakdown of glucose into smaller metabolites. It is widely used in electrochemical glucose sensors designed for diabetes patients.

Features and Benefits

  • Linear detection range: 0.02 to 10 U/L glucose oxidase for the colorimetric assay and 0.002 to 1.5 U/L for the fluorometric assay.

Suitability

The kit is suitable for determining glucose oxidase activity in cell lysate, culture medium and other biological samples, as well as for studying the effects of drugs on glucose metabolism.

Principle

The Glucose Oxidase Assay Kit uses a single working reagent that combines the glucose oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to glucose oxidase activity in the sample.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Glucose oxidase activity measured via continuous spectrophotometric assay at 500 nm, indicating glucose oxidation rate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service