Skip to Content
Merck
All Photos(4)

Key Documents

204005

Sigma-Aldrich

Ammonium dihydrogenphosphate

99.999% trace metals basis

Synonym(s):

Ammonium phosphate monobasic, mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

grade

for analytical purposes

Assay

99.999% trace metals basis

form

crystalline

impurities

≤15.0 ppm Trace Metal Analysis

mp

190 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium dihydrogenphosphate is a white crystalline solid widely applied in rechargeable batteries, non-linear optics, and fire-retardant coatings. It is insoluble in ethanol and acetone.

Application

Ammonium dihydrogenphosphate can be used:
  • As a precursor to fabricate cathode materials for Li-ion batteries to enhance cyclic performance.
  • As an electrolyte additive in rechargeable batteries to enhance the ionic conductivity or to reduce the risk of side reactions within the electrolyte.
  • To fabricate flame-retardant polymer composites.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric S Simon et al.
Analytical biochemistry, 377(2), 234-242 (2008-04-09)
Improvements to phosphopeptide enrichment protocols employing titanium dioxide (TiO2) are described and applied to identification of phosphorylation sites on recombinant human cyclin-dependent kinase 2 (CDK2). Titanium dioxide binds phosphopeptides under acidic conditions, and they can be eluted under basic conditions.
Preparation and electrochemical performance of carbon-coated LiFePO4/LiMnPO4-positive material for a Li-ion battery
Qiangqiang Tan, et al.
Particuology, 30, 144-150 (2017)
Electrochemical properties of a ceramic-polymer-composite-solid electrolyte for Li-ion batteries
Seoung Soo Lee, et al.
Solid State Ionics, 284, 20-24 (2016)
The evaluation of the interfacial and flame retardant properties of glass fiber/unsaturated polyester composites with ammonium dihydrogen phosphate
Jong-Hyun Kim, et al.
Composites Part B: Engineering, 167, 221-230 (2019)
Marija Vukomanovic et al.
Journal of nanobiotechnology, 17(1), 21-21 (2019-02-03)
Emerging concepts for designing innovative drugs (i.e., novel generations of antimicrobials) frequently include nanostructures, new materials, and nanoparticles (NPs). Along with numerous advantages, NPs bring limitations, partly because they can limit the analytical techniques used for their biological and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service