Merck
  • Role of carboxylate chelating agents on the chemical, structural and textural properties of hydroxyapatite.
Role of carboxylate chelating agents on the chemical, structural and textural properties of hydroxyapatite.
Dalton transactions (Cambridge, England : 2003) (2010-10-05)
Karima Achelhi, Sylvie Masse, Guillaume Laurent, Ahmed Saoiabi, Abdelaziz Laghzizil, Thibaud Coradin
ABSTRACT
Organically-modified hydroxyapatite materials were synthesized through the addition of oxalic, succinic, adipic and citric acids to a calcium hydroxide solution before neutralization by ammonium dihydrogenphosphate. All carboxylic acids have a significant influence on apatite crystallinity and nanoparticle size, as indicated by XRD and TEM. Chemical and thermogravimetric analyses as well as FTIR and {(1)H}-(13)C CP MAS NMR spectroscopies indicate that the additives are present in the final material. (1)H, {(1)H}-(31)P HPDec MAS, CP MAS and 2D {(1)H}-(31)P CP-HETCOR MAS NMR experiments suggest that carboxylic acids are localized on the apatite nanocrystallite surface, resulting in the formation of a disordered outer layer. Nitrogen sorption measurements indicate minor modifications of the specific surface area of the resulting mesoporous materials upon carboxylic acid addition but more significant variations in the average dimensions of the pores as well as in the chemical nature of the pore surface. Although these evolutions are mainly in good agreement with the ligand affinity for calcium ions in solution, an unexpected difference was observed between succinic and adipic acid, that may be attributed to steric constraints resulting from the interfacial nature of the calcium-ligand interactions. These data should provide useful guidelines to identify novel efficient additives to control apatite growth.
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammonium phosphate monobasic, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium phosphate dibasic, SAJ first grade, 98.5-101.0%
Sigma-Aldrich
Ammonium phosphate dibasic, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium phosphate monobasic, SAJ first grade, ≥98.0%
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)