Silane A 174 is Methacryloxypropyl trimethoxysilane which should be non-ionic.
1.07673
Silane A 174
adhesion promoter
Synonym(s):
Silane A 174, Polyfix® 100, Methacryloxypropyl trimethoxysilane
About This Item
Recommended Products
vapor pressure
0.023 hPa ( 25 °C)
Quality Level
Assay
≥97% (GC)
form
liquid
potency
>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
mp
<-20 °C
transition temp
flash point 108 °C
density
1.04 g/cm3 at 20 °C
storage temp.
15-25°C
SMILES string
[Si](OC)(OC)(OC)CCCOC(=O)C(=C)C
InChI
1S/C10H20O5Si/c1-9(2)10(11)15-7-6-8-16(12-3,13-4)14-5/h1,6-8H2,2-5H3
InChI key
XDLMVUHYZWKMMD-UHFFFAOYSA-N
Application
- as a silane coupling agent to treat cellulose nanocrystals (CNCs)
- as an adhesion layer/promoter prior to deposition of Parylene C for magnetic microelectromechanical systems (MEMS) fabrication
- to bind the polyacrylamide gels to the glass plate for silver staining
Analysis Note
Identity (IR-spectrum): passes test
Water (according to Karl Fischer): ≤ 0.01 %
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
197.6 °F - closed cup
Flash Point(C)
92 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.
Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.
-
Is A 174 silane anionic or cationic
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service