Skip to Content
Merck
All Photos(1)

Documents

440574

Sigma-Aldrich

Triethoxy(ethyl)silane

96%

Synonym(s):

Ethyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5Si(OC2H5)3
CAS Number:
Molecular Weight:
192.33
Beilstein:
1362028
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

158-160 °C (lit.)

density

0.895 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](CC)(OCC)OCC

InChI

1S/C8H20O3Si/c1-5-9-12(8-4,10-6-2)11-7-3/h5-8H2,1-4H3

InChI key

DENFJSAFJTVPJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

109.9 °F - closed cup

Flash Point(C)

43.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Barbara Vörös-Horváth et al.
Molecules (Basel, Switzerland), 25(23) (2020-12-02)
Onychomycosis is a disease that affects many adults, whose treatment includes both oral and topical therapies with low cure rates. The topical therapy is less effective but causes fewer side effects. This is why the development of an effective, easy
Viktória Lilla Balázs et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-14)
Essential oils (EOs) are becoming increasingly popular in medical applications because of their antimicrobial effect. Direct bioautography (DB) combined with thin layer chromatography (TLC) is a screening method for the detection of antimicrobial compounds in plant extracts, for example, in
Juan A Allegretto et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(54), 12388-12396 (2020-07-17)
This work reports on a novel and versatile approach to control the structure of metal-organic framework (MOFs) films by using polymeric brushes as 3D primers, suitable for triggering heterogeneous MOF nucleation. As a proof-of-concept, this work explores the use of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service