Skip to Content
Merck
All Photos(2)

Key Documents

850725P

Avanti

18:1 (Δ9-Cis) PE (DOPE)

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine; DOPE; PE(18:1(9Z)/18:1(9Z)); 110635, DOPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H78NO8P
CAS Number:
Molecular Weight:
744.03
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850725P-1g)
pkg of 2 × 100 mg (850725P-200mg)
pkg of 5 × 100 mg (850725P-500mg)
pkg of 1 × 25 mg (850725P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C41H78NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,39H,3-16,21-38,42H2,1-2H3,(H,45,46)/b19-17-,20-18-/t39-/m1/s1

InChI key

MWRBNPKJOOWZPW-NYVOMTAGSA-N

General description

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (18:1 (Δ9-Cis) PE (DOPE)) is a neutral phospholipid and displays sensitivity towards pH change. At acidic it is an inverted hexagonal micelle and transforms to spherical micelle at alkaline pH.

Application

18:1 (Δ9-Cis) PE (DOPE) has been used in the preparation of liposome for human uncoupling protein 1 (UCP1) reconstitution and for Caenorhabditis elegans autophagy related cysteine protease protein conjugation studies. It has also been used for the generation of giant unilamellar vesicles (GUVs) for septin induced deformation studies.

Biochem/physiol Actions

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (18:1 (Δ9-Cis) PE (DOPE)) forms a stable lipid layer and rapidly fuses with endosomal lipid bilayer. DOPE in combination with 1,2-dioleoyl-3-trimethylammonium-propane (DOTAP) has high in vitro transfection efficiency.

Packaging

20 mL Clear Glass Screw Cap Vial (850725P-1g)
5 mL Clear Glass Sealed Ampule (850725P-200mg)
5 mL Clear Glass Sealed Ampule (850725P-25mg)
5 mL Clear Glass Sealed Ampule (850725P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring the correlation between lipid packaging in lipoplexes and their transfection efficacy
Moghaddam B, et al.
Pharmaceutics, 3(4), 848-864 (2011)
Autophagy impairment mediated by S-nitrosation of ATG4B leads to neurotoxicity in response to hyperglycemia
Li Y, et al.
Autophagy, 13(7), 1145-1160 (2017)
The role of the helper lipid dioleoylphosphatidylethanolamine (DOPE) for DNA transfection cooperating with a cationic lipid bearing ethylenediamine
Mochizuki S, et al.
Biochimica et Biophysica Acta - Biomembranes, 1828(2), 412-418 (2013)
Specific interaction of the human mitochondrial uncoupling protein 1 with free long-chain fatty acid
Zhao L, et al.
Structure, 25(9), 1371-1379 (2017)
The role of the helper lipid on the DNA transfection efficiency of lipopolyplex formulations
Du Z, et al.
Scientific Reports, 4(2), 7107-7107 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service