Skip to Content
Merck
All Photos(2)

Key Documents

850745P

Avanti

14:0 PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-ditetradecanoyl-sn-glycero-3-phosphoethanolamine; DMPE; PE(14:0/14:0); 110632

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H66NO8P
CAS Number:
Molecular Weight:
635.85
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

description

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (850745P-1g)
pkg of 1 × 200 mg (850745P-200mg)
pkg of 1 × 25 mg (850745P-25mg)
pkg of 1 × 500 mg (850745P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O

InChI

1S/C33H66NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-32(35)39-29-31(30-41-43(37,38)40-28-27-34)42-33(36)26-24-22-20-18-16-14-12-10-8-6-4-2/h31H,3-30,34H2,1-2H3,(H,37,38)/t31-/m1/s1

InChI key

NEZDNQCXEZDCBI-WJOKGBTCSA-N

General description

Phosphatidylethanolamine (PE) is an abundant phospholipid, found in the cells of mammals.

Application

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine (14:0 PE):
  • has been used as a phospholipid (PL) internal standard in hydrophilic interaction liquid chromatography tandem mass spectrometry (HILIC-LC-MS/MS)
  • has been used as an internal lipid standard to correct the reproducibility in matrix-assisted laser desorption/ionization Fourier transform ion cyclotron resonance mass spectrometry (MALDI FT-ICR MS) analysis
  • may be used in proteoliposomes for planar supported lipid bilayer formation
  • may be used as a biotinylated internal standard to measure externalization of aminophospholipids (APLs) using mass spectrometry (MS)

Biochem/physiol Actions

Phosphatidylethanolamine (PE) plays a key role in the activity of respiratory complexes. PE also participates in the initiation of autophagy.

Packaging

20 mL Clear Glass Screw Cap Vial (850745P-1g)
20 mL Clear Glass Screw Cap Vial (850745P-500mg)
5 mL Amber Glass Screw Cap Vial (850745P-200mg)
5 mL Amber Glass Screw Cap Vial (850745P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex J B Kreutzberger et al.
Biophysical journal, 115(9), 1755-1761 (2018-10-22)
Intramembrane proteases hydrolyze peptide bonds within the membrane as a regulatory paradigm that is conserved across all forms of cellular life. Many of these enzymes are thought to be oligomeric, and that their resulting quaternary interactions form the basis of
Paulina Perczyk et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183136-183136 (2019-11-22)
Soil fungi play an important role in the environment decomposing dead organic matter and degrading persistent organic pollutants (POP). The presence of hydrophobic POP in the soil and membrane-lytic substances excreted by competing microorganism to the soil solution is the
Paulina Perczyk et al.
Biochimica et biophysica acta. Biomembranes, 1862(6), 183239-183239 (2020-03-03)
Soil bacteria are decomposer organisms crucial for the biodegradation of organic pollutants, mineralization of dead organic matter and the turnover of biogenic elements. In their environment they are constantly exposed to membrane-lytic enzymes emitted to the soil by other microorganisms
Erum Malik et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183141-183141 (2019-12-04)
Linearized esculentin 2 EM (E2EM-lin) from the frog, Glandirana emeljanovi was highly active against Gram-positive bacteria (minimum lethal concentration ≤ 5.0 μM) and strongly α-helical in the presence of lipid mimics of their membranes (>55.0%). The N-terminal α-helical structure adopted by E2EM-lin showed
Tomas Laursen et al.
Science (New York, N.Y.), 354(6314), 890-893 (2016-11-20)
Metabolic highways may be orchestrated by the assembly of sequential enzymes into protein complexes, or metabolons, to facilitate efficient channeling of intermediates and to prevent undesired metabolic cross-talk while maintaining metabolic flexibility. Here we report the isolation of the dynamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service