Skip to Content
Merck
All Photos(2)

Key Documents

810145P

Avanti

18:1 NBD PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H82N5O11P
CAS Number:
Molecular Weight:
924.15
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810145P-1mg)
pkg of 5 × 1 mg (810145P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

18:1 NBD PE is a fluorescently labelled lipid, containing unsaturated C18:1Δ9-acyl chains.

Application

18:1 NBD PE is suitable for:
  • preparing liposomes
  • fluorescence spectra study
  • labelling giant unilamellar vesicles

Biochem/physiol Actions

L-α-Phosphatidylethanolamine (PE) is essential for maintaining the structural integrity of membranes.

Packaging

5 mL Amber Glass Screw Cap Vial (810145P-1mg)
5 mL Amber Glass Screw Cap Vial (810145P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assembling responsive microgels at responsive lipid membranes
Wang M, et al.
Proceedings of the National Academy of Sciences of the USA, 116(12), 5442-5450 (2019)
Fusion of single proteoliposomes with planar, cushioned bilayers in microfluidic flow cells
Karatekin E and Rothman JE
Nature Protocols, 7(5), 903-903 (2012)
A Janus-faced IM30 ring involved in thylakoid membrane fusion is assembled from IM30 tetramers
Saur M, et al.
Structure, 25(9), 1380-1390 (2017)
Black Lipid Membranes: Challenges in Simultaneous Quantitative Characterization by Electrophysiology and Fluorescence Microscopy
Tsemperouli M, et al.
Langmuir (2019)
Felista L Tansi et al.
Pharmaceutics, 12(4) (2020-04-23)
Liposomes are biocompatible nanocarriers with promising features for targeted delivery of contrast agents and drugs into the tumor microenvironment, for imaging and therapy purposes. Liposome-based simultaneous targeting of tumor associated fibroblast and the vasculature is promising, but the heterogeneity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service